Key Documents
55085-U
Upchurch In-Line Cartridge Check Valve
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.
Related Categories
General description
The inline Cartridge Check Valve is designed to limit flow to one direction. The cartridge and flangeless fitting is for use with 1/16″ O.D. tubing. The internal volume of the check valve is less than 150μL. Place in the flow system where you wish to restrict fluid flow to one direction. These assemblies withstand system pressures of 1.000 psi (69 bar). Complete with wrench.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)