54283-U
Supel™ Sphere Carbon/NH2 SPE Tube
volume 6 mL, pk of 30
Synonym(s):
Supel™ Sphere
About This Item
Recommended Products
material
polypropylene tube
Quality Level
product line
Supel™
composition
bed A, 500 mg (Spherical carbon)
bed B, 500 mg (Spherical aminopropyl (NH2) phase)
packaging
pk of 30
technique(s)
solid phase extraction (SPE): suitable
volume
6 mL
matrix active group
amino phase
carbon phase
application(s)
food and beverages
General description
Sample Matrix Compatibility: Organic or aqueous solutions
- Dual layer SPE tube that contains both spherical carbon (upper layer) and spherical aminopropyl (NH2) phase (lower layer) SPE sorbents, separated by PE frit.
- Developed to offer superior cleanup when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.).
- Carbon has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products.
- Aminopropyl (NH2) retains fatty acids, organic acids, and some polar pigments and sugars common in food matrices.
- SPE tube packed entirely with spherical, non-friable particles
- Improved flow characteristics and faster flow for gravity filtration
- Reduced susceptibility to the formation of fines
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service