Skip to Content
Merck
All Photos(1)

Key Documents

48731

Supelco

Aroclor 1242 solution

certified reference material, 500 mg/kg in transformer oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 5 mL

concentration

500 mg/kg in transformer oil

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

room temp

SMILES string

Clc1c(ccc(c1)Cl)c2c(cc(cc2)Cl)Cl

InChI

1S/C12H6Cl4/c13-7-1-3-9(11(15)5-7)10-4-2-8(14)6-12(10)16/h1-6H

InChI key

QORAVNMWUNPXAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ashley Smithwick et al.
Toxicology, 204(1), 61-74 (2004-09-17)
Immunological effects of polychlorinated biphenyls (PCBs) have been demonstrated in our laboratories with the peferential inhibition of lipopolysaccharide (LPS)-induced splenocyte proliferation by ortho-substituted PCB congeners. An investigation of the mechanism behind this immunotoxicity revealed an interruption in the progression of
L Ashley Smithwick et al.
Toxicology, 188(2-3), 319-333 (2003-05-28)
Polychlorinated biphenyls (PCBs) are persistent environmental contaminants, and their ubiquitous nature has prompted studies of their potential health hazards. As a result of their lipophilic nature, PCBs accumulate in breast milk and subsequently affect the health of offspring of exposed
Syed Uzair Ahmad et al.
Environmental research, 93(3), 272-278 (2003-11-15)
Adult male rhesus monkeys (Macaca mulatta) were given oral treatment of either Aroclor 1242 or vehicle (corn oil and glycerol) at a dose of 200 microg/kg body wt/day for 6 months to investigate the effects of the pollutant on plasma
Na Ding et al.
FEMS microbiology ecology, 70(2), 149-158 (2009-08-12)
Polychlorinated biphenyls in a commercial mixture (Aroclor 1242) were added to soil at 8.0 mg kg(-1) with and without ryegrass (Lolium multiflorum L.) planted in a specially designed rhizobox. At the end of 90 days, the presence of plants significantly
Valentina Tremaroli et al.
FEMS microbiology ecology, 74(2), 291-301 (2010-09-18)
Pseudomonas pseudoalcaligenes KF707 is a polychlorinated biphenyls (PCBs) degrader, also tolerant to several toxic metals and metalloids. The work presented here examines for the first time the chemotactic response of P. pseudoalcaligenes KF707 to biphenyl and intermediates of the PCB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service