Skip to Content
Merck
All Photos(1)

Key Documents

47994

Supelco

Bis(2-ethylhexyl) phthalate solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 506

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

O(CC(CCCC)CC)C(=O)c1c(cccc1)C(=O)OCC(CCCC)CC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here
Bis(2-ethylhexyl) phthalate belongs to the class of phthalate esters, released in the environment through volatilization and leaching from plastics and other sources. It is extensively used as an additive for the manufacturing of polyvinyl chloride (PVC) plastics.

Application

Bis(2-ethylhexyl) phthalate may be used as a reference standard for the quantification of the analyte in milk samples and water samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a headspace solid-phase microextraction method combined with gas chromatography mass spectrometry for the determination of phthalate esters in cow milk
Feng L-Y, et al.
Analytica Chimica Acta, 538(1-2), 41-48 (2005)
Development of a solid-phase microextraction method for the determination of phthalic acid esters in water.
Prokupkova G, et al.
Analytica Chimica Acta, 457(2), 211-223 (2002)
Pinar Erkekoglu et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 31(2), 143-153 (2012-12-12)
This study was designed to investigate the effects of di-(2-ethylhexyl) phthalate (DEHP) on thyroid hormone levels and oxidant/antioxidant parameters in the rat and to evaluate the effects of selenium status. Selenium deficiency was produced by feeding 3-week-old Sprague-Dawley rats with
Xue-Feng Huang et al.
PloS one, 7(11), e50465-e50465 (2012-12-12)
The present study was undertaken to determine the reproductive hazards of Di-(2-ethylhexyl)-phthalate (DEHP) on mouse spermatozoa and embryos in vitro and genomic changes in vivo. Direct low-level DEHP exposure (1 μg/ml) on spermatozoa and embryos was investigated by in vitro
Mingyong Li et al.
Environmental toxicology and pharmacology, 35(1), 55-60 (2012-12-12)
The aim of this study was to evaluate dose-related effects on external genitalia of adult male offspring rats by maternal exposure to di-(2-ethylhexyl) phthalate (DEHP) plasticizer. Timed-pregnant rats were given DEHP by gastric intubation at doses of 0, 500, 750

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service