Skip to Content
Merck
All Photos(1)

Key Documents

46951-U

Supelco

trans-9,12-Octadecadienoic acid methyl ester

certified reference material, 10 mg/mL in heptane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

Assay

99.9% (chromatography)

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

single component solution

functional group

ester (methyl)

storage temp.

-10 to -25°C

SMILES string

O(C)C(=O)CCCCCCC\C=C\C\C=C\CCCCC

InChI

1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11H,3-6,9,12-18H2,1-2H3/b8-7+,11-10+

InChI key

WTTJVINHCBCLGX-ZDVGBALWSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Roman Shchepin et al.
Journal of the American Chemical Society, 132(49), 17490-17500 (2010-11-26)
Free radical co-oxidation of polyunsaturated lipids with tyrosine or phenolic analogues of tyrosine gave rise to lipid peroxide-tyrosine (phenol) adducts in both aqueous micellar and organic solutions. The novel adducts were isolated and characterized by 1D and 2D NMR spectroscopy
Akio Kadowaki et al.
Chemistry and physics of lipids, 165(2), 178-185 (2012-01-03)
The antioxidative action of fullerenes has received much attention, but their reaction mechanism toward lipid-derived peroxyl radicals has not been well elucidated. In this study, the reaction products of [60]fullerene (C(60)) during the autoxidation of methyl linoleate (MeL) were isolated
L Tesoriere et al.
Free radical research, 43(8), 706-717 (2009-06-24)
Reaction kinetics of betanin and its aglycone betanidin towards peroxyl radicals generated from the azo-initiated oxidation of methyl linoleate in methanol and of a heterogeneous aqueous/soybean phosphatidylcholine liposomal system were studied by monitoring formation of linoleic acid hydroperoxides and consumption
Yasutaka Shimotori et al.
Carbohydrate research, 359, 11-17 (2012-08-29)
Various glycosyl ferulates were efficiently synthesized from 2,4,6-tetra-O-acetyl-α-D-glucopyranosyl bromide (TAGB) with amine by amine-promoted glycosylation without using heavy metal. The resulted acetylated glycosyl ferulates with acetoxyl groups at C-2, C-3 and C-4 were regioselectively deacetylated at C-4 and C-6 positions
Philip T Lynett et al.
Organic & biomolecular chemistry, 9(9), 3320-3330 (2011-03-30)
It has long been recognized that garlic and petiveria, two plants of the Allium genus--which also includes onions, leeks and shallots--possess great medicinal value. In recent times, the biological activities of extracts of these plants have been ascribed to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service