Skip to Content
Merck
All Photos(1)

Key Documents

40542

Supelco

Aniline solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Nc1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

PAYRUJLWNCNPSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Blood, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conghui Tang et al.
Journal of the American Chemical Society, 134(46), 18924-18927 (2012-11-08)
A novel and efficient copper-catalyzed azidation reaction of anilines via C-H activation has been developed. This method, in which the primary amine acts as a directing group by coordinating to the metal center, provides ortho azidation products regioselectively under mild
Sung Jin Bae et al.
European journal of medicinal chemistry, 57, 383-390 (2012-11-15)
We attempted to design and synthesize (E)-N-substituted benzylidene-hydroxy or methoxy-aniline derivatives and to evaluate their inhibitory effect on tyrosinase activity and anti-melanogenesis activity in murine B16F10 melanoma cells. Derivatives with a 4-methoxy- or 4-hydroxy-anilino group exerted more potent inhibition against
Huihui Yang et al.
Electrophoresis, 34(4), 510-517 (2012-11-23)
A novel (3-sulfopropyl methacrylate potassium)-silica hybrid monolithic column for CEC has been prepared by a simple one-pot approach based on efficient thiol-ene click chemistry. In this process, the polycondensation of hydrolyzed alkoxysilanes and in situ click reaction of vinyl groups
Laya Siroos Rezaei et al.
Environmental technology, 33(19-21), 2273-2280 (2013-02-12)
Aerobic granules can be formed in sequencing batch airlift reactors (SBAR) and sequencing batch reactors (SBR). Comparing these two systems, the SBAR has excellent mixing condition, but due to a high height-to-diameter ratio (H/D), there is no performance capability at
Ping-Xin Zhou et al.
Chemical communications (Cambridge, England), 49(31), 3254-3256 (2013-03-14)
Isoindolines are synthesized by palladium-catalyzed coupling reaction of N-(2-iodobenzyl) anilines with α,β-unsaturated N-tosylhydrazones. The reaction has several potential advantages: (1) toleration of a wide range of functional groups, (2) easy to handle and with mild conditions, (3) enriches the isoindoline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service