27136
Vials, screw top with phenolic open-top cap, pre-assembled
volume 4 mL, clear glass vial, O.D. × H 15 mm × 45 mm, tan PTFE/silicone septum
All Photos(1)
About This Item
material
black phenolic cap (open-top)
clear glass vial
tan PTFE/silicone septum
feature
closure type black phenolic hole cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
15 mm × 45 mm
volume
4 mL
fitting
thread for 13-425
Application
Glass screw top vials (40 ml) sealed with phenolic open-top caps and PTFE/silicone septa may be used to contain pectin or plant material, during δ2H analysis of plant methoxyl groups, in an experimental study to analyse stable hydrogen-isotope of methyl chloride, which is emitted from heated halophytic plants.
Choose from one of the most recent versions:
Certificates of Analysis (COA)