Skip to Content
Merck
All Photos(1)

Documents

24565

Supelco

Supelcarb HC Hydrocarbon Trap

volume 750 cc, 1/2 in. fitting

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

volume

750 cc

Quality Level

fitting

1/2 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

Hydrocarbon traps are basically used in GC for carrier gas purification, as the name suggests they are primarily used for trapping hydrocarbons.
Supelcarb adsorbent has the greatest trapping ability of hydrocarbons per gram of adsorbent (twice that of activated charcoal). Simply stated, Supelcarb HC hydrocarbon traps are the best trap available for removing hydrocarbons and other organics from carrier, fuel, air, and other gas streams.

Dimensions:
  • 120 cc Trap: 111/8in. (28.2 cm) long x 13/8 in. (35 mm) diameter
  • 750 cc Trap: 161/2 in. (41.9 cm) long x 25/16in. (59 mm) diameter

Legal Information

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Smith-Cortinez et al.
Cells, 9(11) (2020-11-15)
Upon liver injury, hepatic stellate cells (HSCs) transdifferentiate to migratory, proliferative and extracellular matrix-producing myofibroblasts (e.g., activated HSCs; aHSCs) causing liver fibrosis. HSC activation is associated with increased glycolysis and glutaminolysis. Here, we compared the contribution of glycolysis, glutaminolysis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service