Skip to Content
Merck
All Photos(1)

Key Documents

20811

Supelco

ORBO 80 Isocyanate Filter

with cassettes: no, filter O.D. 37 mm, pkg of 25 ea

Synonym(s):

ORBO 80 Coated Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

material

glass fiber filter

Agency

ASTM® D5836
OSHA 5002

product line

ORBO

packaging

pkg of 25 ea

manufacturer/tradename

ORBO 80

technique(s)

active air sampling: suitable

filter O.D.

37 mm

matrix active group

1,2-PP, 1-(2-pyridyl)piperazine

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for analyte group isocyanate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

ORBO 80 Coated 1,2-PP Filter is a 1-(2-pyridyl) piperazine coated glass fiber filter best suited for sampling diisocyanates in the air (OSHA Methods 42 and 47).

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bjørn T Moe et al.
Anticancer research, 29(4), 1047-1052 (2009-05-06)
Endometrial hyperplasia is a precursor lesion of endometrial carcinoma. Clinical studies of endometrial hyperplasia have shown that levonorgestrel (LNG) is more therapeutically effective than medroxyprogesterone acetate (MPA). The present pharmacological in vitro study was performed to compare progestin effects on
A Orbo et al.
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke, 117(7), 972-976 (1997-03-10)
The development of female steroid hormone-related neoplasms such as gynaecologic and mammary cancers is influenced by hormones administered exogenously. Hormonal contraceptives and hormone replacement therapy both affect the risk of developing cancers in the endometrium, cervix, ovarium, and mammary glands.
K Flo et al.
Scandinavian journal of clinical and laboratory investigation, 55(8), 715-721 (1995-12-01)
Elevated extracellular cGMP levels have been observed in various clinical conditions, and the analyte has been proposed as a diagnostic marker of cardiovascular as well as malignant diseases. However, the use of extracellular cGMP as a pathophysiological marker requires detailed
Anne Ørbo et al.
The Journal of steroid biochemistry and molecular biology, 113(1-2), 139-149 (2009-01-14)
Patients with endometrial hyperplasia representing preliminary stages of endometrial cancer have shown to respond to therapy in 100% of the cases when treated with levonorgestrel-impregnated intrauterine device. Anti-proliferative effect has also been reported after application of an anti-progestin impregnated intrauterine
A Orbo et al.
International journal of oncology, 5(3), 619-625 (1994-09-01)
About 50% of the cases with invasive carcinoma of the uterine cervix occurs before menopause. Cyclic changes of sex steroids may therefore interfere with the ratio between extracellular levels of cGMP and cAMP (cGMP(ex)/cAMP(ex)). This ratio has been proposed as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service