Skip to Content
Merck
All Photos(1)

Key Documents

11809

Supelco

Coated GC Packing

phase 1% SP®-1510, matrix 60/80 Carbopack B support, bottle of 15 g

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23151817

packaging

bottle of 15 g

Quality Level

parameter

60-230 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix

60/80 Carbopack B support

matrix active group

1% SP®-1510 phase

column type

packed GC

compatibility

for analyte group Industrial solvents

Looking for similar products? Visit Product Comparison Guide

General description

The choice of coating material is based on the stationary phase of the GC. Carbopack Adsorbent can withstand high bake-out temperature without losing the mechanical and chemical integrity. It has a high affinity for isoprene and does not retain significant amounts of water. Furthermore it has a tendency to pulverize if it is packed too tightly.
Ready-to-use coated packing for those wanting to pack their own columns.

Application

Carbopack B was used for the analysis of acetaldehyde, methanol, and ethanol by headspace gas chromatography. It was also used for the analysis of phenylurea herbicides in environmental aqueous samples.

Legal Information

Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
SP is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and sensitive determination of phenylurea herbicides in water in the presence of their anilines by extraction with a Carbopack cartridge followed by liquid chromatography.
di Corcia A and Marchetti M.
Journal of Chromatography A, 541, 365-373 (1991)
Romy Gleyse Chagas Barros et al.
Food research international (Ottawa, Ont.), 130, 108899-108899 (2020-03-12)
Edible flowers are receiving renewed interest as potential sources of bioactive compounds. The present study aimed to investigate the presence of bioactive compounds and antioxidant activity of some exotic flowers present in Brazil such as Amaranthus hypochondriacus, Tropaeolum majus (red)
Analytical Measurements in Aquatic Environments.
Namiesnik J and Szefer P.
Science, 73-73 (2009)
R M Anthony et al.
Journal of analytical toxicology, 4(1), 43-45 (1980-01-01)
This report describes a headspace procedure suitable for the gas chromatographic assay of acetaldehyde, methanol, ethanol and various other volatile substances in biological samples. Carbopack B coated with 5% Carbowax 20-M is compared to Carbopack C coated with 0.2% Carbowax
Plant Volatile Analysis.
Linskens HF and Jackson JF.
Modern Methods of Plant Analysis, 19, 54-56 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service