Skip to Content
Merck
All Photos(1)

Key Documents

10284

HayeSep® Porous Polymer Adsorbent

matrix HayeSep A, 100-120 mesh, bottle of 75 cc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
eCl@ss:
32119290

form

solid

packaging

bottle of 75 cc

manufacturer/tradename

Hayes Separation Inc

parameter

165 °C temp. limit

technique(s)

gas chromatography (GC): suitable

surface area

~526 m2/g

matrix

HayeSep A

particle size

100-120 mesh

density

~0.36 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

HayeSep porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. HayeSep® has a high surface area owing to its micropores thereby making it a suitable candidate for separation of gases and volatile organic compounds (VOCs). Furthermore they are relatively inert and exhibit hydrophobicity. HayeSep®A is a copolymer of divinylbenzene and ethylene glycol methacrylate.

Application

HayeSep®A maybe used for the measurement of carbon dioxide using the on-line gas analyser during the experimental set-up, to use activated carbon as a buffer in biofiltration of waste gases, to ensure a constant supply of contaminants like toluene to the bioreactor for effective biological waste-gas treatment.

Legal Information

HayeSep is a registered trademark of Hayes Separation Inc.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas Chromatography in Air Pollution Analysis
Berezkin V.G and Drugov Y.S. et al.
J. Chromatogr. Library, 49, 50-51 (1991)
Use of activated carbon as a buffer in biofiltration of waste gases with fluctuating concentrations of toluene.
Weber FJ and Hartmans S.
Applied Microbiology and Biotechnology, 43 (2), 365-369 (1995)
Gas Chromatography
Poole C. et al.
Science, 127-130 (2012)
Trace Analysis of Specialty and Electronic Gases.
Geiger W.M and Raynor M.W. et al.
Science, 256-257 (2013)
Specific features of the gas-chromatographic determination of the reaction products in the catalytic decomposition of hydrogen sulfide.
Zheivot V, et al.
Journal of Analytical Chemistry, 61 (4), 334-337 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service