Skip to Content
Merck
All Photos(9)

Key Documents

ZRB5320

Sigma-Aldrich

Anti-NG2 Antibody, clone 1L2, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Chondroitin sulfate proteoglycan 4, Chondroitin sulfate proteoglycan NG2, HSN tumor-specific antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L2, monoclonal
recombinant monoclonal

description

1L2 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 251.91 kDa
observed mol wt ~270 kDa

species reactivity

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunofluorescence: suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1L2 is a ZooMab rabbit recombinant monoclonal antibody that specifically detects Chondroitin sulfate proteoglycan 4 (NG2).

Immunogen

Recombinant extracellular domain of rat Chondroitin sulfate proteoglycan 4 (NG2).

Application

Anti-NG2, clone 1L2 ZooMAb, Cat. No. ZRB5320, is a recombinant Rabbit Monoclonal Antibody that specifically targets Chondroitin sulfate proteoglycan 4 (NG2) and has been tested for use in Immunocytochemistry, Immunofluorescence, and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected NG2 in C6 cells.

Immunofluorescence Analysis: A 1:100 dilution representative lot detected NG2 in rat brain tissue sections.

Target description

Chondroitin sulfate proteoglycan 4 (UniProt: Q00657; also known as Chondroitin sulfate proteoglycan NG2, HSN tumor-specific antigen) is encoded by the Cspg4 (also known as Ng2) gene (Gene ID: 81651) in rat. Chondroitin sulfate proteoglycan NG2 is a single-pass type I membrane protein that plays a role in cell proliferation and migration and stimulates endothelial cells motility during microvascular morphogenesis. It is present in neural cells and in extraneural tissues, especially in the developing mesenchyme. Its level of expression is highest on immature, proliferating cells and decreases when these cells begin to differentiate. It is found on the surfaces of an unusual class of glial cells within the developing and mature central nervous system that have the properties of oligodendrocyte precursor cells. It is a major chondroitin sulfate proteoglycan that is produced after spinal cord injury and is expressed by macrophages and oligodendrocyte progenitors. It is also reported to inhibit neurite outgrowth and growth cone collapse during axon regeneration. NG2 chondroitin sulfate proteoglycan is synthesized with a signal peptide (aa 1-29), which is subsequently cleaved off in the mature form. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Jones, LL et al. (2002). J. Neurosci. 22(7):2792-2803).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Tedeschi et al.
PloS one, 17(4), e0267576-e0267576 (2022-04-28)
Retinopathy of prematurity (ROP) remains the leading cause for blindness in children. Limited hyperoxia induced proliferative retinopathy (L-HIPR) was recently introduced as a potential animal model for ROP and persistent fetal vasculature; however, the detailed pathological changes remain unclear. To
Huitong Song et al.
Glia, 69(12), 2863-2881 (2021-08-17)
Therapeutics that promote oligodendrocyte survival and remyelination are needed to restore neurological function in demyelinating diseases. Sphingosine 1-phosphate (S1P) is an essential lipid metabolite that signals through five G-protein coupled receptors. S1P receptor agonists such as Fingolimod are valuable immunosuppressants
Nikolett Nagy et al.
Scientific reports, 14(1), 13139-13139 (2024-06-08)
The enzyme dUTPase has an essential role in maintaining genomic integrity. In mouse, nuclear and mitochondrial isoforms of the enzyme have been described. Here we present the isoform-specific mRNA expression levels in different murine organs during development using RT-qPCR. In

Related Content

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service