Z741651
Sartorius pipette tips
volume range 500-10000 μL, Standard, bulk, non-sterile
All Photos(2)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
1 mL
volume range:
500-10000 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
endotoxin free
RNase free
barrier: no
endotoxin free
packaging:
pack of 100 ea
manufacturer/tradename:
Sartorius 780310
Recommended Products
material
colorless
colorless tip
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
endotoxin free
packaging
pack of 100 ea
manufacturer/tradename
Sartorius 780310
maximum volume
1 mL
volume range
500-10000 μL
General description
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Optifit Tips are available in various packaging types for all your needs:
- Single Tray racks – purity certified tip racks
- Refill Packs - purity certified and ecological refill packages
- Refill Towers - space-saving and ecological refill packages
- Flexibulk boxes - space-saving, ecological and economical bulk boxes
- Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)