Z328723
BRAND® magnetic stirring bar, PTFE, cylindrical
L 15 mm, bar diam. 6 mm
All Photos(1)
About This Item
UNSPSC Code:
41103806
eCl@ss:
32100100
NACRES:
NB.72
Recommended Products
material
PTFE-coated
cylindrical
manufacturer/tradename
BRAND 137114
L
15 mm
bar diam.
6 mm
Related Categories
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
- BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
- The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
- Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137114 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)