Documents
WHAWB100014
QIAcard Bloodstain non-indicating FTA cards
pack of 100 cards
Synonym(s):
QIAcard Bloodstain (100), Whatman FTA, Z719838, blood stain cards
About This Item
UNSPSC Code:
41105500
NACRES:
NB.22
manufacturer/tradename
Qiagen WB100014
packaging
pack of 100 cards
suitability
suitable for (collection and transport of blood and bodily fluids)
General description
The QIAcard Bloodstain (4 sample areas per card) is a non-Indicating and non-FTA treated card designed for the collection and short-term handling of blood and bodily fluids. Unlike FTA Cards, the Bloodstain Cards are not coated with a protective chemical cocktail and DNA is as risk of degradation and damage. Convenient transport of blood and bodily fluids with protective cover included.
Features and Benefits
  • Designed for the collection and transport of blood and bodily fluids
  • Each card features a protective cover
  • Blood stain cards are designed for short-term handling of specimens
  • Card enables efficient sample collection
  • 125 μL sample per spot
Other Notes
Field of Use : For internal research use only
Legal Information
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Visit the Document Library
Protocols
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.