Skip to Content
Merck
All Photos(1)

Key Documents

W4016

Sigma-Aldrich

Wedelolactone

≥98% (HPLC), powder

Synonym(s):

1,8,9-Trihydroxy-3-methoxy-6H-benzofuro[3,2-c][1]benzopyran-6-one, 7-Methoxy-5,11,12-trihydroxycoumestan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10O7
CAS Number:
Molecular Weight:
314.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

COc1cc(O)c-2c(OC(=O)c3c-2oc4cc(O)c(O)cc34)c1

InChI

1S/C16H10O7/c1-21-6-2-10(19)14-12(3-6)23-16(20)13-7-4-8(17)9(18)5-11(7)22-15(13)14/h2-5,17-19H,1H3

InChI key

XQDCKJKKMFWXGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Wedelolactone inhibits NF-κB-mediated gene transcription in cells by blocking the phosphorylation and degradation of IκBα. Irreversible inhibitor of IKKα and β kinase activity (IC50 < 10 μM). Wedelolactone has no effects on p38 MAP kinase or Akt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi-Mei Liu et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(11), 4016-4022 (2012-08-21)
Eclipta prostrata L., (Asteraceae), is used in China for both food and medicine purposes. This research is concerned with the isolation and purification of phytochemical constituents from the aerial parts of E. prostrata, using gradient solvent fractionation, macroporous resin, silica
Kumiko Tanabe et al.
Journal of neuroinflammation, 7, 16-16 (2010-03-09)
Interleukin (IL)-6 plays a pivotal role in a variety of CNS functions such as the induction and modulation of reactive astrogliosis, pathological inflammatory responses and neuroprotection. Tumor necrosis factor (TNF)-alpha induces IL-6 release from rat C6 glioma cells through the
Kumiko Tanabe et al.
Journal of neuroinflammation, 8, 68-68 (2011-06-21)
Peripheral-type benzodiazepine receptor (PBR) expression levels are low in normal human brain, but their levels increase in inflammation, brain injury, neurodegenerative states and gliomas. It has been reported that PBR functions as an immunomodulator. The mechanisms of action of midazolam
Petr Benes et al.
Cancer letters, 303(1), 29-38 (2011-02-15)
The naturally occurring coumestan wedelolactone has been previously shown to reduce growth of various cancer cells. So far, the growth-suppressing effect of wedelolactone has been attributed to the inhibition of the NFκB transcription factor and/or androgen receptors. We found that
Zhimin Chen et al.
The Journal of biological chemistry, 288(20), 14417-14427 (2013-04-13)
Signal transducers and activators of transcription 1 (STAT1) transduces signals from cytokines and growth factors, particularly IFN-γ, and regulates expression of genes involved in cell survival/death, proliferation, and migration. STAT1 is activated through phosphorylation on its tyrosine 701 by JAKs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service