Key Documents
V8255
Supelco
Liquid scintillation vials, glass
volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with metal foil liner. (caps packed separately), pkg of 500 ea
Synonym(s):
Glass Vials for Scintillation Counting
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
material
glass vial
metal foil liner
polypropylene cap
feature
closure type screw top vial
closure type white urea cap
packaging
pkg of 500 ea
vial size
28 mm × 61 mm , with cap
volume
20 mL
fitting
thread for 22-400
General description
Wheaton 180 brand low-potassium glass yielding consistent low backgrounds and high UV transmission. White caps fit tightly and can be written on.
Packaging
Vials are shrink-wrapped in individual cell trays of 100.
Caps are packed in sacks of 100 in separate trays.