Skip to Content
Merck
All Photos(1)

Key Documents

V4388

Sigma-Aldrich

Anti-VP16 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti Alpha trans-inducing protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

immunoprecipitation (IP): 10 μg using mammalian cell extracts expressing VP16 fusion proteins
western blot: 1:1,000 using mammalian cell extracts expressing VP16 fusion proteins

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-VP16 is produced in rabbit using a synthetic peptide corresponding to amino acids 474-487 of the herpes simplex virus VP16 protein, conjugated to KLH via an N-terminal added cysteine residue. Whole antiserum is fractionated and further purified by ionexchange chromatography to provide the IgG fraction of antiserum that is essentially free of other rabbit serum proteins.

Immunogen

synthetic peptide corresponding to amino acids 474-487 of the herpes simplex virus VP16 protein, conjugated to KLH via an N-terminal added cysteine residue.

Application

Anti-VP16 recognizes VP16 fusion proteins by immunoblotting and immunoprecipitation. It is used to study the effect of sialic acid on herpes simplex virus type 1 envelope glycoproteins. It is also used to study if self-association of lymphocytic choriomeningitis virus nucleoprotein is mediated by its N-terminal region and is not required for its anti-interferon function.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeremy R Teuton et al.
Journal of virology, 81(8), 3731-3739 (2007-01-19)
Herpes simplex virus type 1 (HSV-1) envelope proteins are posttranslationally modified by the addition of sialic acids to the termini of the glycan side chains. Although gC, gD, and gH are sialylated, it is not known whether sialic acids on
Ying-Hsiu Su et al.
Journal of virology, 80(23), 11589-11597 (2006-09-22)
Following infection, the physical state of linear herpes simplex virus (HSV) genomes may change into an "endless" or circular form. In this study, using Southern blot analysis of the HSV genome, we provide evidence that immediate-early protein ICP4 is involved
Health care's future.
R J Nowak
Ohio medicine : journal of the Ohio State Medical Association, 87(5), 229-230 (1991-05-01)
Ida G Lunde et al.
The Journal of physiology, 582(Pt 3), 1277-1287 (2007-04-28)
The effects of exercise on skeletal muscle are mediated by a coupling between muscle electrical activity and gene expression. Several activity correlates, such as intracellular Ca(2+), hypoxia and metabolites like free fatty acids (FFAs), might initiate signalling pathways regulating fibre-type-specific
Jamie Snider et al.
Nature protocols, 5(7), 1281-1293 (2010-07-03)
The biological function of proteins may be predicted by identification of their interacting partners, and one of the major goals of the postgenomic era is the mapping of protein interaction networks. Membrane proteins are of particular interest because of their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service