Skip to Content
Merck
All Photos(2)

Key Documents

V2889

Sigma-Aldrich

Valeryl Salicylate

≥98% (HPLC)

Synonym(s):

2-Valeryloxybenzoic acid, Valerylsalicylic acid, vsa

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

mp

86-87 °C (lit.)

solubility

DMSO: 24 mL/mL
H2O: insoluble

SMILES string

CCCCC(=O)Oc1ccccc1C(O)=O

InChI

1S/C12H14O4/c1-2-3-8-11(13)16-10-7-5-4-6-9(10)12(14)15/h4-7H,2-3,8H2,1H3,(H,14,15)

InChI key

WJHZBTMHUNVIKC-UHFFFAOYSA-N

Biochem/physiol Actions

Selective, irreversible COX-1 inhibitor. The selectivity for COX-1 vs COX-2 shows the IC50 values for ovine COX-1 vs COX-2 of 0.8 μM vs. 15 mM respectively.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharath Kandhi et al.
Pulmonary circulation, 9(4), 2045894019895947-2045894019895947 (2020-01-08)
Epoxyeicosatrienoic acids (EETs) are synthesized from arachidonic acid by CYP/epoxygenase and metabolized by soluble epoxide hydrolase (sEH). Roles of EETs in hypoxia-induced pulmonary hypertension (HPH) remain elusive. The present study aimed to investigate the underlying mechanisms, by which EETs potentiate
Eduardo Candelario-Jalil et al.
Journal of neuroinflammation, 4, 25-25 (2007-10-12)
Neuroinflammatory responses are triggered by diverse ethiologies and can provide either beneficial or harmful results. Microglial cells are the major cell type involved in neuroinflammation, releasing several mediators, which contribute to the neuronal demise in several diseases including cerebral ischemia
Jing Huang et al.
Pulmonary circulation, 9(1), 2045894019837876-2045894019837876 (2019-02-27)
Caveolin-1 (cav-1) has been shown to play a significant role in the pathogenesis of pulmonary hypertension (PH). In the monocrotaline model of PH, the loss of endothelial cav-1 as well as reciprocal activation of proliferative and anti-apoptotic pathways initiate the
Hui Chen et al.
American journal of physiology. Heart and circulatory physiology, 314(3), H681-H691 (2017-11-12)
Hypertension is associated with endothelial dysfunction, which favors the release of endothelium-derived contracting factors, including vasoconstrictor prostanoids and reactive oxygen species. Peroxisome proliferator-activated receptor-α (PPAR-α) agonists, clinically used as lipid-lowering drugs, possess antioxidant properties and exert beneficial effects in the
Ju-Fang Liu et al.
Molecular cancer, 9, 43-43 (2010-02-25)
Cyclooxygenase (COX)-2, the inducible isoform of prostaglandin (PG) synthase, has been implicated in tumor metastasis. Interaction of COX-2 with its specific EP receptors on the surface of cancer cells has been reported to induce cancer invasion. However, the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service