Skip to Content
Merck
All Photos(4)

Key Documents

U2625

Sigma-Aldrich

Uric acid

≥99%, crystalline

Synonym(s):

2,6,8-Trihydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O3
CAS Number:
Molecular Weight:
168.11
Beilstein:
156158
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.55
Pricing and availability is not currently available.

Quality Level

100
200

Assay

≥99%

form

crystalline

mp

>300 °C (lit.)

SMILES string

O=C1NC(=O)C2=C(N1)NC(=O)N2

InChI

1S/C5H4N4O3/c10-3-1-2(7-4(11)6-1)8-5(12)9-3/h(H4,6,7,8,9,10,11,12)

InChI key

LEHOTFFKMJEONL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Uric acid has been used:
  • in inducing hyperuricemia in mice[1]
  • as a component of digestion fluid for the simulation of in vitro human digestion model[2]
  • in selectivity test of the glucose sensor in sweat-based diabetes monitoring [3]

Biochem/physiol Actions

Uric acid is an insoluble catabolite produced by adenine and guanine metabolism. Accumulation of uric acid leads to gout, hyperuricemia, arthritis and renal failure.[4] Elevated uric acid levels contributes to hypertension and pathogenesis of cardiovascular disease.[5][6] Low uric acid levels associated with Parkinson′s disease and multiple sclerosis, may elicit protective functionality.[6] High levels of uric acid in patients with chronic obstructive pulmonary disease (COPD) may serve as potential marker for diagnosis.[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A graphene-based electrochemical device with thermoresponsive microneedles for diabetes monitoring and therapy
Lee H, et al.
Nature Nanotechnology, 11(6), 566-566 (2016)
Serum uric acid: a risk factor and a target for treatment?
Feig DI, et al.
Journal of the American Society of Nephrology, 17(4), S69-S73 (2006)
Uric acid in plants and microorganisms: Biological applications and genetics-A review
Hafez RM, et al.
Journal of Advanced Research, 8(5), 475-486 (2017)
Reduction of Serum Uric Acid Associated with Attenuation of Renal Injury, Inflammation and Macrophages M1/M2 Ratio in Hyperuricemic Mice Model
HARYONO A, et al.
Kobe J. Med. Sci., 64(3), E107-E114 (2018)
Uric acid, hyperuricemia and vascular diseases
Jin M, et al.
Frontiers in Bioscience, 17, 656-656 (2012)

Articles

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service