Skip to Content
Merck
All Photos(2)

Documents

U103

Sigma-Aldrich

U-69593

solid

Synonym(s):

(+)-(5α,7α,8β)-N-Methyl-N-[7-(1-pyrrolidinyl)-1-oxaspiro[4.5]dec-8-yl]-benzeneacetamide, U69593

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32N2O2
CAS Number:
Molecular Weight:
356.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

optical activity

[α]/D +7.8°, c = 0.825 in methanol(lit.)

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 10 mg/mL
0.1 M HCl: >40 mg/mL
ethanol: >40 mg/mL
0.1 M NaOH: insoluble
H2O: insoluble

storage temp.

2-8°C

SMILES string

CN([C@H]1CC[C@@]2(CCCO2)C[C@@H]1N3CCCC3)C(=O)Cc4ccccc4

InChI

1S/C22H32N2O2/c1-23(21(25)16-18-8-3-2-4-9-18)19-10-12-22(11-7-15-26-22)17-20(19)24-13-5-6-14-24/h2-4,8-9,19-20H,5-7,10-17H2,1H3/t19-,20-,22-/m0/s1

InChI key

PGZRDDYTKFZSFR-ONTIZHBOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

U-69593 is a selective κ opioid receptor agonist. U-69593 is known to inhibit cocaine sensitization in meso-limbic dopamine neurons by normalizing basal overflow of dopamine.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

U-69593 is soluble in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin (10 mg/ml), 0.1 M HCl (>40 mg/ml), and ethanol (>40 mg/ml). However, it is insoluble in 0.1 M NaOH and water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmel M McDermott et al.
The Journal of physiology, 589(Pt 14), 3517-3532 (2011-05-25)
The dentate gyrus of the hippocampus is thought to control information flow into the rest of the hippocampus. Under pathological conditions, such as epilepsy, this protective feature is circumvented and uninhibited activity flows throughout the hippocampus. Many factors can modulate
C A Heidbreder et al.
Neuroreport, 5(14), 1797-1800 (1994-09-08)
Repeated intermittent administration of cocaine (20 mg kg-1, i.p.) for 3 days dramatically increased basal dopamine (DA) overflow in the nucleus accumbens (ACB) 48 h after the final daily injection. This cocaine pretreatment also produced a significant increase in stereotypy
S Stevens Negus et al.
Psychopharmacology, 210(2), 149-159 (2010-01-27)
Selective, centrally acting kappa opioid agonists produce antinociception in a wide range of preclinical assays, but these compounds perform poorly as analgesics in humans. This discrepancy may be related to the behavioral depressant effects of kappa agonists. Kappa antagonists do
Gregory P McLennan et al.
Journal of neurochemistry, 107(6), 1753-1765 (2008-11-19)
GTP binding regulatory protein (G protein)-coupled receptors can activate MAPK pathways via G protein-dependent and -independent mechanisms. However, the physiological outcomes correlated with the cellular signaling events are not as well characterized. In this study, we examine the involvement of
S Stevens Negus et al.
Experimental and clinical psychopharmacology, 16(5), 386-399 (2008-10-08)
Micro opioid receptor agonists are clinically valuable as analgesics; however, their use is limited by high abuse liability. Kappa opioid agonists also produce antinociception, but they do not produce micro agonist-like abuse-related effects, suggesting that they may enhance the antinociceptive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service