Key Documents
TMO382019-0030
Nalgene® Certified Clean bottles
clear PETG, volume 30 mL, case of 72 ea
Synonym(s):
PETG bottle, certified nalgene bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
clear PETG
packaging
case of 72 ea
manufacturer/tradename
Nalgene 382019-0030
capacity
30 mL
volume
30 mL
General description
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Features and Benefits
  • Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)