Key Documents
TMO362006-9025
Nalgene® narrow-mouth bottle without cap
amber polypropylene copolymer, volume 8 mL, case of 2000 ea
Synonym(s):
Nalgene bottle, plastic bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
amber polypropylene copolymer
packaging
case of 2000 ea
manufacturer/tradename
Nalgene 362006-9025
capacity
8 mL
volume
8 mL
General description
Safely store and handle test kits and many small volume applications with Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)