Skip to Content
Merck
All Photos(3)

Key Documents

T6451

Sigma-Aldrich

Anti-Twist1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Twist homolog 1 (acrocephalosyndactyly 3, Saethre-Chotzen syndrome), Anti-Twist related protein 1 (H-twist)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~26 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 1-2 μg/mL using human osteosarcoma MG-63 cells
western blot (chemiluminescent): 0.5-1 μg/mL using human 293T cells expressing recombinant human Twist1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TWIST1(7291)
mouse ... Twist1(22160)
rat ... Twist1(85489)

General description

Twist is encoded by the gene mapped to human chromosome 7p21.1. The encoded protein is a basic helix-loop-helix (bHLH) transcription factor. Twist 1 belongs to the Twist gene family and is mainly expressed in the suture mesenchyme.

Specificity

Anti-Twist1 antibody recognizes human Twist1. Detection of the Twist1 band by immunoblotting is specifically inhibited with the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 12-27 of human Twist1 with a C-terminal added cysteine, conjugated to KLH. The corresponding peptide sequence is identical in both rat and mouse.

Application

Anti-Twist1 antibody is suitable for use in detecting Twist1 via in immunochemical applications such as immunoblotting (~26 kDa) and immunofluorescence.
Immunoblotting: a working antibody concentration of 0.5-1 mg/mL is recommended using extracts of human 293T cells expressing recombinant human Twist1 and a chemiluminescent detection reagent.
Anti-Twist1 antibody produced in rabbit has been used in:
  • immunohistochemistry
  • immunofluorescence
  • immunoblotting
  • chromatin immunoprecipitation (ChIP)

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Biochem/physiol Actions

Twist is required for mesoderm formation. Twist 1 controls embryonic morphogenesis. It also plays a key role in metastasis by inducing an epithelial-mesenchymal transition (EMT). During an EMT, Twist stimulates the expression of mesenchymal markers, such as fibronectin and N-cadherin. In addition, it also blocks E-cadherin expression inhibiting cell-cell adhesion and therefore inducing infiltrative tumor growth in several human carcinomas. Twist1 is required in head mesenchyme for cranial neural tube morphogenesis in mice. Twist1 is involved in osteoblast differentiation and maturation. Increased expression of Twist 1 in breast cancer cells induce angiogenesis and leads to chromosomal instability. Mutations in this gene have been found in patients with Saethre-Chotzen syndrome, an autosomal dominant defect characterized by minor skull and limb anomalies.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Twist1 directly regulates genes that promote cell proliferation and migration in developing heart valves
Lee MP and Yutzey KE
PLoS ONE, 6(12), e29758-e29758 (2011)
Twist, a Master Regulator of Morphogenesis, Plays an Essential Role in Tumor Metastasis
Yang J, et al.
Cell, 117 (2004)
Jonathan M Mudry et al.
The Journal of endocrinology, 224(3), 303-313 (2015-02-11)
TWIST proteins are important for development of embryonic skeletal muscle and play a role in the metabolism of tumor and white adipose tissue. The impact of TWIST on metabolism in skeletal muscle is incompletely studied. Our aim was to assess
Tori E Horne et al.
Journal of cardiovascular development and disease, 2(3), 214-232 (2015-11-04)
The heart valve interstitial cell (VIC) population is dynamic and thought to mediate lay down and maintenance of the tri-laminar extracellular matrix (ECM) structure within the developing and mature valve throughout life. Disturbances in the contribution and distribution of valve
Lu Wang et al.
The EMBO journal, 36(20), 2987-2997 (2017-09-09)
DNA methylation is a major epigenetic modification; however, the precise role of DNA methylation in vertebrate development is still not fully understood. Here, we show that DNA methylation is essential for the establishment of the left-right (LR) asymmetric body plan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service