Skip to Content
Merck
All Photos(1)

Documents

T5574

Sigma-Aldrich

Terrific Broth

Liquid microbial growth medium

Synonym(s):

TB medium, Tartoff – Hobbs medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

for molecular biology

description

Liquid Medium

sterility

sterile; 0.2 μm filtered

form

liquid

technique(s)

microbiological culture: suitable

application(s)

microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Terrific Broth is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains glycerol, peptides, amino acids, water-soluble vitamins, and carbohydrates.

Application

Terrific Broth has been used for non-selective cultivation of E. coli strains for cloning, high-yield DNA plasmid production, and high-yield production of recombinant proteins. citation It is also suitable for selective cultivation when appropriate antibiotics are added.

Features and Benefits

Terrific Broth liquid provides:
  • Filter-sterilized, ready-to-use format
  • Convenient package size
  • Standard formulation

Components

23.6g/L Yeast Extract
11.8g/L Tryptone
9.4g/L K2HPO4
2.2g/L KH2PO4
4ml/L Glycerol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaheen Shojania et al.
Journal of virological methods, 164(1-2), 35-42 (2009-11-28)
The HIV-1 transactivator of transcription (Tat) is a protein essential for virus replication. Tat is an intrinsically disordered RNA-binding protein that, in cooperation with host cell factors cyclin T1 and cyclin-dependent kinase 9, regulates transcription at the level of elongation.
Shani Leviatan et al.
The Journal of biological chemistry, 285(31), 23548-23556 (2010-06-08)
Membrane proteins constitute 20-30% of all proteins encoded by the genome of various organisms. Large amounts of purified proteins are required for activity and crystallization attempts. Thus, there is an unmet need for a heterologous membrane protein overexpression system for
Andrey Sokolov et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(3), 969-974 (2010-01-19)
Whereas the laws of thermodynamics prohibit extraction of useful work from the Brownian motion of particles in equilibrium, these motions can be "rectified" under nonequilibrium conditions, for example, in the presence of asymmetric geometrical obstacles. Here, we describe a class
The preparation and analysis of DNA for use in transgenic technology.
Auerbach, A.B., et al.
Advanced Protocols For Animal Transgenesis, 501-501 (2011)
Ørjan Samuelsen et al.
Antimicrobial agents and chemotherapy, 52(8), 2905-2908 (2008-06-19)
Purified recombinant VIM-7 possesses efficient penicillinase and carbapenemase activities comparable to those of VIM-2. Cephalosporinase activity was variable and generally lower than those of VIM-1 and VIM-2. A homology model suggests that the VIM-7 Tyr-218 Phe substitution may be responsible

Protocols

Preparation of Plasmid DNA by Alkaline Lysis with SDS: Maxipreparation between Cold Spring Harbor Laboratory Press and our research team.

General protocols for growth of competent cells in microbial medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service