T5300
Transforming Growth Factor-β2 from porcine platelets
≥97% (SDS-PAGE and N-terminal analysis), recombinant, powder, suitable for cell culture
Synonym(s):
TGF-β2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
Transforming Growth Factor-β2 from porcine platelets, powder, suitable for cell culture
biological source
Porcine (Platelets)
Quality Level
Assay
≥97% (SDS-PAGE and N-terminal analysis)
form
powder
potency
0.05-0.5 ng/mL ED50/EC50
mol wt
protein 25 kDa
packaging
pkg of 1 μg
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
pig ... TGFB2(397084)
General description
TGF-β2 (transforming growth factor-β2) belongs to the TGF-β superfamily and the gene is mapped to pig (Sus scrofa) chromosome 10p16.
Biochem/physiol Actions
TGF-β2 is responsible for maintaining immune homeostasis. TGF-β2 promotes cellular growth, differentiation and survival. TGF-β2 controls lymphocyte proliferation, apoptosis, hematopoiesis, and embryogenesis. TGF-β2 is a tumor suppress or in the early stages of carcinogenesis, but in the later stages acts as a tumor promoter by inducing epithelial-mesenchymal transition and stimulating angiogenesis. TGF-β2 prevents NK (natural killer) cells growth as well as B and T cell proliferation. TGF-β2 also greatly inhibits the growth of both normal and transformed epithelial. The growth of lymphoid, fibroblast and keratinocyte cells are also known to be hinder by TGF-β2. TGF-β2 controls intestinal homeostasis by functioning in synergy with the bacterial lipopolysaccharide and thereby maintain interleukin-8.
TGF-β2, like TGF-β1, is produced by many cell types and reported to be most concentrated in mammalian platelets.
TGF-β2 belongs to the TGF-β superfamily. TGF-β2 is important for immune homeostasis by balancing lymphocyte proliferation, apoptosis, hematopoiesis, and embryogenesis. TGF-β2 is crucial in cell growth, differentiation, and survival. TGF-β2 is a strong growth inhibitor for normal and transformed epithelial, lymphoid, fibroblast, and keratinocyte cells. TGF-β2 is a tumor suppress or in the early stages of carcinogenesis, but in the later stages acts as a tumor promoter by inducing epithelial-mesenchymal transition and stimulating angiogenesis. TGF-β2 inhibits NK cells growth as well as B and T cell proliferation.
TGF-β2 belongs to the TGF-β superfamily. TGF-β2 is important for immune homeostasis by balancing lymphocyte proliferation, apoptosis, hematopoiesis, and embryogenesis. TGF-β2 is crucial in cell growth, differentiation, and survival. TGF-β2 is a strong growth inhibitor for normal and transformed epithelial, lymphoid, fibroblast, and keratinocyte cells. TGF-β2 is a tumor suppress or in the early stages of carcinogenesis, but in the later stages acts as a tumor promoter by inducing epithelial-mesenchymal transition and stimulating angiogenesis. TGF-β2 inhibits NK cells growth as well as B and T cell proliferation.
TGF-β2, like TGF-β1, is produced by many cell types and reported to be most concentrated in mammalian platelets.
Physical form
Lyophilized from a 0.2 μm filtered solution in 25% acetonitrile and 0.1% trifluoroacetic acid containing 0.05 mg bovine serum albumin.
Analysis Note
The biological activity is measured by its ability to inhibit the IL-4-dependent proliferation of mouse HT-2 cells.
The biological activity of porcine TGF-β2 was tested in culture by measuring its ability to inhibit thymidine incorporation in the IL-4 dependent mouse T-helper cell line HT-2. The EC50 is defined as the effective concentration of growth factor that elicits 50% inhibition of cell growth in a cell based bioassay.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Signaling Pathway Involving TGF-β2 and Snail in Hair Follicle Morphogenesis
PLoS Biology, 3.1 (2004)
Transforming growth factor-? induces epithelial to mesenchymal transition and suppresses the proliferation and transdifferentiation of cultured human pancreatic duct cells.
Journal of Cellular Biochemistry (2010)
Transforming growth factor-2 and endotoxin interact to regulate homeostasis via interleukin-8 levels in the immature intestine
American Journal of Physiology: Gastrointestinal and Liver Physiology, 307, G689- G699 (2014)
Mapping of the porcine urate oxidase and transforming growth factor β 2 genes by fluorescence in situ hybridization.
Chromosome Research, 4(2), 147-150 (1996)
Curcumin suppresses doxorubicin-induced epithelial?mesenchymal transition via the inhibition of TGF-β and PI3K/AKT signaling pathways in triple-negative breast cancer cells.
Journal of Agricultural and Food Chemistry, 61(48), 11817-11824 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service