Skip to Content
Merck
All Photos(1)

Key Documents

T4077

Sigma-Aldrich

Anti-TAZ antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Taz Antibody, Taz Antibody - Anti-TAZ antibody produced in rabbit, Anti-Transcriptional co-activator with PDZ-binding motif

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~49 kDa

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... WWTR1(25937)

Related Categories

General description

Tafazzin (TAZ) is encoded by the WW domain containing transcription regulator 1 (WWTR1) gene mapped to human chromosome Xq28. The encoded protein contains 400 amino acids. It is characterized by a conserved WW domain, implicated in binding PPXY motif, a coiled-coil region associated with protein-protein interaction and a C-terminal motif involved in binding the PDZ domain.

Immunogen

synthetic peptide corresponding to residues 386-400 of human TAZ.

Biochem/physiol Actions

Tafazzin (TAZ) plays a vital role in the various cellular process including cell proliferation, differentiation, apoptosis, migration, invasion, epithelial-mesenchymal transition (EMT) and stemness in multiple human cancers. TAZ associates with other transcription factors such as RUNX2 (runt-related transcription factor 2), PPAR (peroxisome proliferator-activated receptor), PAX3 and 8 (paired box gene 3 and 8) and TTF1 (thyroid transcription factor 1) and plays an essential role in osteoblastic, myogenic and adipogenic differentiation.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristen M Stearns-Reider et al.
Aging cell, 16(3), 518-528 (2017-04-04)
Age-related declines in skeletal muscle regeneration have been attributed to muscle stem cell (MuSC) dysfunction. Aged MuSCs display a fibrogenic conversion, leading to fibrosis and impaired recovery after injury. Although studies have demonstrated the influence of in vitro substrate characteristics on
Claudio Pulito et al.
Journal of experimental & clinical cancer research : CR, 38(1), 349-349 (2019-08-11)
Over the past decade, newly designed cancer therapies have not significantly improved the survival of patients diagnosed with Malignant Pleural Mesothelioma (MPM). Among a limited number of genes that are frequently mutated in MPM several of them encode proteins that
Intra-individual plasticity of the TAZ gene leading to different heritable mutations in siblings with Barth syndrome.
Ferri L
European Journal of Human Genetics, 23, 1708-1712 (2015)
Regulation of TAZ in cancer.
Zhou X and Lei QY
Protein & cell, 7, 548-561 (2016)
Shu Li et al.
Apoptosis : an international journal on programmed cell death, 29(7-8), 1198-1210 (2024-03-30)
A number of studies have confirmed that Yes-associated protein (YAP)/transcriptional co-activator with PDZ-binding motif (TAZ)-transcriptional enhanced associate domain (TEAD) activity is the driver of cancer development. However, the role and mechanism of the YAP/TAZ-TEAD pathway in cervical intraepithelial neoplasia (CIN)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service