Skip to Content
Merck
All Photos(1)

Key Documents

T3634

Sigma-Aldrich

(+)-α-Tocopherol

from vegetable oil, Type V, ~1000 IU/g

Synonym(s):

2,5,7,8-Tetramethyl-2-(4′,8′,12′-trimethyltridecyl)-6-chromanol, 5,7,8-Trimethyltocol, D-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
4712525
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058016
PubChem Substance ID:
NACRES:
NA.79

biological source

vegetable oil

type

Type V

form

viscous liquid

specific activity

~1000 IU/g

color

clear yellow to red

refractive index

n20/D 1.505 (lit.)

bp

200-220 °C/0.1 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29-/m1/s1

InChI key

GVJHHUAWPYXKBD-IEOSBIPESA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Tocopherol is synthesized from γ-tocopherol by the action of enzyme γ-tocopherol methyltransferase. It is the major form of Vitamin E in human plasma. It is present in sunflower seed oil.

Application

(+)-α-Tocopherol has been used:
  • as a food supplement for juvenile cobia fish
  • to evaluate its protective effect on bisphenol A induced oxidative stress in rats
  • to test its chemopreventive efficacy in estrogen-mediated mammary cancer rats

Biochem/physiol Actions

α-Tocopherol is essential for the photosynthesis in Synechocystis sp. strain PCC 6803. Supplementation with α-Tocopherol decreases lipid peroxidation and platelet aggregation. It inhibits protein kinase C and may play key role in gene regulation.
Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens. Various isofroms and analogues of tocopherol have opposing and differentiated regulatory activities in vivo.

Packaging

Packaged under argon in a sealed ampule.

Other Notes

Mixed constitutional (α, β, γ, δ) isomers.

Quality

Approx. 670 mg D-α-tocopherol per gram. The non-α content is typically between 5-35 mg/g

Caution

Subject to air oxidation.

Physical properties

This product is miscible with chloroform or ethanol. It is practically insoluble in water but it is miscible with ether, acetone, and vegetable oils. It is unstable to alkaline conditions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of alpha-tocopherol and alpha-lipoic acid on bisphenol-A-induced oxidative damage in liver and ovarian tissue of rats
Avci B, et al.
Toxicology and Industrial Health, 32(8), 1381-1390 (2016)
Taro Sakamoto et al.
Journal of clinical biochemistry and nutrition, 66(2), 116-123 (2020-04-02)
Glutathione peroxidase 4 (GPx4) is a unique antioxidant enzyme that directly reduces the phospholipid hydroperoxides (PLOOH) generated in biomembranes using glutathione as the reductant. We have previously reported that the Caenorhabditis elegans gpx-quad mutant, which lacks all homologous genes of
Dietary alanyl-glutamine and vitamin E supplements could considerably promote the expression of GPx and PPARalpha genes, antioxidation, feed utilization, growth, and improve composition of juvenile cobia
Ding Z, et al.
Aquaculture (Amsterdam, Netherlands), 470(6), 95-102 (2017)
alpha-Tocopherol is essential for acquired chill-light tolerance in the cyanobacterium Synechocystis sp. strain PCC 6803
Yang Y, et al.
Journal of Bacteriology, 190(5), 1554-1560 (2008)
The role of alpha-tocopherol in preventing disease: from epidemiology to molecular events
Azzi A, et al.
Molecular Aspects of Medicine, 24(6), 325-336 (2003)

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service