Skip to Content
Merck
All Photos(6)

Key Documents

T2885

Sigma-Aldrich

3,3′,5,5′-Tetramethylbenzidine

TMB substrate, chromogenic, ≥98% (TLC), powder

Synonym(s):

BM blue, Sure Blue TMB, TMB, TMB Blotting Plus, TMB substrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H2(CH3)2-4-NH2]2
CAS Number:
Molecular Weight:
240.34
Beilstein:
2808541
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

mp

168-171 °C (lit.)

solubility

ethyl acetate: 50 mg/mL, clear to very slightly hazy

storage temp.

2-8°C

SMILES string

Cc1cc(cc(C)c1N)-c2cc(C)c(N)c(C)c2

InChI

1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3

InChI key

UAIUNKRWKOVEES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3′,5,5′-Tetramethylbenzidine (TMB) is a noncarcinogenic substitute (Ames test negative) for benzidine suitable as peroxidase substrate for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases.
3,3′,5,5′-Tetramethylbenzidine has been used:
  • as a supplement in Man-Rodosa-Sharpe (MRS) agar plate, to stains the lactobacilli colonies blue in the presence of O2
  • as a substrate solution for ELISA (enzyme-linked immunosorbent assay)
  • in myeloperoxidase activity assay to quantify neutrophil

Biochem/physiol Actions

3,3′,5,5′-Tetramethylbenzidine/TMB can be used as a chromogen to increase the progression of product obtained in peroxidase reaction. In food and environmental decontamination procedures, TMB can be used in in situ free available chlorine (FAC) monitoring of chlorite-based sanitizers.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferrotransferrin and antibody against the transferrin receptor as potential vehicles for drug delivery across the mammalian blood-brain barrier into the central nervous system
Methods in Neurosciences, 21(1), 93-117 (1994)
Lactobacilli isolated from vaginal vault of dairy and meat cows during progesteronic stage of estrous cycle
Rodriguez C, et al.
Anaerobe, 17(1), 15-18 (2011)
Tetramethylbenzidine method for monitoring the free available chlorine and microbicidal activity of chlorite-based sanitizers under organic-matter-rich environments
Yamaoka H, et al.
Letters in Applied Microbiology, 62(1), 47-54 (2016)
Lactobacillus casei improves resistance to pneumococcal respiratory infection in malnourished mice
Villena J, et al.
The Journal of Nutrition, 135(6), 1462-1469 (2005)
The CD4+ T cell-mediated IFN-gamma response to Helicobacter infection is essential for clearance and determines gastric cancer risk
Sayi A, et al.
Journal of immunology (Baltimore, Md. : 1950), 182(11), 7085-7101 (2009)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service