Skip to Content
Merck

T2391

Triosephosphate Isomerase from rabbit muscle

Type III-S, ammonium sulfate suspension, ≥4,000 units/mg protein

Synonym(s):

D-Glyceraldehyde-3-phosphate ketol-isomerase, TPI

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
≥4,000 units/mg protein
Biological source:
rabbit muscle
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit muscle

Quality Segment

type

Type III-S

form

ammonium sulfate suspension

specific activity

≥4,000 units/mg protein

mol wt

calculated mol wt 53.2 kDa

foreign activity

3-phosphoglyceric phosphokinase ≤0.01%, glyceraldehyde-3-phosphate dehydrogenase ≤0.01%, pyruvate kinase, lactic dehydrogenase, phosphoglucose isomerase, α-glycerophosphate dehydrogenase and aldolase ≤0.001%

storage temp.

2-8°C

Application

Triosephosphate isomerase has been used in a study to assess molecular characterizations of Cryptosporidium, Giardia, and Enterocytozoon. Triosephosphate isomerase has also been used in a study to investigate apoptosis of hepatocellular carcinoma cell lines.

Physical form

Crystalline suspension in 3.2 M (NH4)2SO4 solution, pH 6.0

Analysis Note

Protein determined by biuret

Other Notes

One unit will convert 1.0 μmole D-glyceraldehyde 3-phosphate to dihydroxyacetone phosphate per min at pH 7.6 at 25 °C.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library