T1943
Greiner centrifuge tubes
centrifuge tube, 15 mL, 17x120 mm, conical (V) bottom, w/ graduation, I.D. field
Synonym(s):
centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, Greiner centrifuge tubes
About This Item
material
cap
conical bottom tube
polypropylene tube
Agency
suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010
reg. compliance
suitable for FDA C-010.02
sterility
sterile
feature
RCF 3,500 × g
graduations, ID field
packaging
pack of 100
case of 1000
manufacturer/tradename
Greiner 188271-N
capacity
15 mL
diam. × L
17 mm × 120 mm
color
blue cap
colorless tube
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
General description
High quality, crystal-clear polypropylene centrifuge tubes. Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service