Skip to Content
Merck
All Photos(4)

Documents

T1912

Sigma-Aldrich

Paclitaxel

from Taxus yannanensis, powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
Beilstein:
1420457
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

biological source

Taxus yannanensis

Quality Level

form

powder

color

white

mp

213 °C (dec.) (lit.)

solubility

DMSO: 50 mg/mL (can be stored frozen for several months)
acetonitrile: soluble
ethanol: soluble
methanol: soluble (undergoes transesterification)

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes

originator

Bristol-Myers Squibb

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](O)[C@@]3(C)C(=O)[C@H](OC(C)=O)C4=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)c5ccccc5)[C@]3([H])[C@@]1(CO2)OC(C)=O)C4(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7

InChI

1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32-,33+,35-,36+,37+,38-,40-,45+,46-,47+/m0/s1

InChI key

RCINICONZNJXQF-MZXODVADSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: taxoide

Application

Paclitaxel has been used to study PREP2-tubulin interactions using coimmunoprecipitation assays on NIH3T3 cell extracts. This drug has also been used to stabilize tubulin obtained from pig brain.

Biochem/physiol Actions

Paclitaxel is a potent anti-neoplastic and anti-mitotic taxane drug, which binds to the N-terminus of β-tubulin and and stabilizes microtubules arresting the cell cycle at the G2/M phase. The microtubule damage induces apoptosis through a JNK-dependent pathway followed by a JNK-independent pathway, perhaps related to the activation of protein kinase A (PKA) or of Raf-1 kinase, resulting in phosphorylation of Bcl-2. A major metabolite via CYP2CB is 6α-hydroxypaclitaxel (6α-OHP).

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

Paclitaxel undergoes transesterification in methanol and hydrolyzes in aqueous solutions.

Preparation Note

Paclitaxel is soluble in DMSO at 50 mg/ml and can be stored frozen for several months. It is also soluble in methanol (undergoes transesterification), acetonitrile and ethanol. Paclitaxel is rapidly destroyed in weakly alkaline, methanolic solutions and in strongly acidic methanolic solutions (1:1 of methanol:concentrated HCl). It is also soluble in a mixture of 50% Cremophor EL and 50% anhydrous ethanol.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1B - STOT RE 1

Target Organs

Central nervous system,Bone marrow,Cardio-vascular system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akane Furuta et al.
The Journal of biological chemistry, 284(9), 5927-5935 (2009-01-07)
Outer arm dynein (OAD) of cilia and flagella contains two or three distinct heavy chains, each having a motor function. To elucidate their functional difference, we compared the in vitro motile properties of Chlamydomonas wild-type OAD containing the alpha, beta
Klaus Haller et al.
The Journal of biological chemistry, 279(47), 49384-49394 (2004-09-02)
The PREP, MEIS, and PBX families are mammalian members of the TALE (three amino acid loop extension) class of homeodomain-containing transcription factors. These factors have been implicated in cooperative DNA binding with the HOX class of homeoproteins, but PREP and
Eric Pujade-Lauraine et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(13), 1302-1308 (2014-03-19)
In platinum-resistant ovarian cancer (OC), single-agent chemotherapy is standard. Bevacizumab is active alone and in combination. AURELIA is the first randomized phase III trial to our knowledge combining bevacizumab with chemotherapy in platinum-resistant OC. Eligible patients had measurable/assessable OC that
George Au-Yeung et al.
Gynecologic oncology, 133(1), 16-22 (2014-04-01)
Obesity is an increasing health problem that is reported to influence chemotherapy dosing. The extent to which this occurs and whether this affects outcomes in ovarian cancer was unclear. To describe chemotherapy dosing practices in normal, overweight and obese patients
Makoto Iimori et al.
Nature communications, 7, 11117-11117 (2016-04-01)
Temporal regulation of microtubule dynamics is essential for proper progression of mitosis and control of microtubule plus-end tracking proteins by phosphorylation is an essential component of this regulation. Here we show that Aurora B and CDK1 phosphorylate microtubule end-binding protein

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service