Skip to Content
Merck
All Photos(1)

Key Documents

T1692

Sigma-Aldrich

Tianeptine sodium salt hydrate

≥98% (HPLC), powder

Synonym(s):

7-[(3-Chloro-6,11-dihydro-6-methyl-dibenzo[c,f][1,2]thiazepine-11-yl)amino]heptanoic acid S,S dioxide sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24ClN2NaO4S · xH2O
CAS Number:
Molecular Weight:
458.93 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥20 mg/mL

originator

Servier

storage temp.

room temp

SMILES string

O.[Na+].CN1c2ccccc2C(NCCCCCCC([O-])=O)c3ccc(Cl)cc3S1(=O)=O

InChI

1S/C21H25ClN2O4S.Na.H2O/c1-24-18-9-6-5-8-16(18)21(23-13-7-3-2-4-10-20(25)26)17-12-11-15(22)14-19(17)29(24,27)28;;/h5-6,8-9,11-12,14,21,23H,2-4,7,10,13H2,1H3,(H,25,26);;1H2/q;+1;/p-1

InChI key

HNTRDGDHBBFLPG-UHFFFAOYSA-M

General description

Tianeptine is a μ-opioid receptor (MOR) as well as a δ-opioid receptor (DOR) agonist.

Application

Tianeptine sodium salt hydrate has been used as a selective serotonin reuptake enhancer (SSRE) positive control during serotonin transporter (SERT) assay in human embryonic kidney 293 cell lines.

Biochem/physiol Actions

Tianeptine is a tricyclic antidepressant that enhances serotonin uptake. This drug can affect neuroprotection, neuronal excitability, memory and anxiety. However, tianeptine is not linked to adverse psychomotor, cognitive, cardiovascular, sleep and bodyweight effects. The drug has a low tendency for abuse and has a positive pharmacokinetic profile.
Tianeptine is least involved in drug interaction as it is not metabolized by cytochrome P450 system in the liver. In hippocampus and amygdala, tianeptine is involved in glutamatergic transmission processes and elicits neuroplasticity.

Features and Benefits

This compound was developed by Servier. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Tianeptine sodium salt hydrate is soluble in DMSO at a concentration that is greater than or equal to 20 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Frankfurt et al.
Brain research, 696(1-2), 1-6 (1995-10-23)
Tianeptine is an atypical tricyclic antidepressant that facilitates serotonin (5-HT) reuptake. Tianeptine (10 mg/kg) or saline was administered intraperitoneally to male rats daily for 4 days. Monoamine levels were measured in micropunches of discrete brain nuclei that are implicated in
W Sohn et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 24(9), 860-e398-860-e398 (2012-06-12)
Tricyclic antidepressants have good efficacy in irritable bowel syndrome with diarrhea (IBS-D), but their clinical use is limited by considerations of tolerability. Tianeptine, another antidepressant, acts as a selective serotonin reuptake enhancer. We compared tianeptine with amitriptyline for the treatment
A Sah et al.
Translational psychiatry, 2, e171-e171 (2012-10-11)
Adult neurogenesis has been implicated in affective disorders and the action of antidepressants (ADs) although the functional significance of this association is still unclear. The use of animal models closely mimicking human comorbid affective and anxiety disorders seen in the
Zorana Oreščanin-Dušić et al.
Acta physiologica Hungarica, 99(2), 140-147 (2012-08-02)
Tianeptine is a novel anti-depressant with an efficacy equivalent to that of classical anti-depressants. Additional beneficial effects include neuroprotection, anti-stress and anti-ulcer properties whose molecular mechanisms are still not completely understood but may involve changes in the anti-oxidant defence system.
C D Conrad et al.
Behavioral neuroscience, 110(6), 1321-1334 (1996-12-01)
Chronic restraint stress causes significant dendritic atrophy of CA3 pyramidal neurons that reverts to baseline within a week. Therefore, the authors assessed the functional consequences of this atrophy quickly (within hours) using the Y maze. Experiments 1-3 demonstrated that rats

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service