Skip to Content
Merck
All Photos(2)

Key Documents

T0656

Sigma-Aldrich

1,3,4,6-Tetrachloro-3α,6α-diphenylglycouril

Synonym(s):

Iodo-Gen®, NSC 4462

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10Cl4N4O2
CAS Number:
Molecular Weight:
432.09
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

storage temp.

2-8°C

Quality Level

SMILES string

ClN1C(=O)N(Cl)C2(N(Cl)C(=O)N(Cl)C12c3ccccc3)c4ccccc4

InChI

1S/C16H10Cl4N4O2/c17-21-13(25)23(19)16(12-9-5-2-6-10-12)15(21,11-7-3-1-4-8-11)22(18)14(26)24(16)20/h1-10H

InChI key

FJQZXCPWAGYPSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Iodination reagent used in radioiodination acting as an oxidative agent

Reagent involved in comparison studies with chloramine-T

Reagent used for oxidation of urazoles

Legal Information

Iodo-Gen is a registered trademark of Pierce Biotechnology, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernard M Tijink et al.
European journal of nuclear medicine and molecular imaging, 36(8), 1235-1244 (2009-03-05)
The human monoclonal antibody (MAb) fragment L19-SIP is directed against extra domain B (ED-B) of fibronectin, a marker of tumour angiogenesis. A clinical radioimmunotherapy (RIT) trial with (131)I-L19-SIP was recently started. In the present study, after GMP production of (124)I
Ulla Lund et al.
American journal of physiology. Renal physiology, 284(6), F1226-F1234 (2003-03-07)
The size and charge-selective properties of the glomerular barrier are partly controversial. Glomerular sieving coefficients (theta) for proteins have rarely been determined noninvasively before in vivo. Therefore, theta was assessed vs. glomerular filtration rate (GFR; (51)Cr-EDTA clearance) in intact rats
Miro J Eigenmann et al.
The Journal of physiology, 595(24), 7311-7330 (2017-09-30)
For therapeutic antibodies, total tissue concentrations are frequently reported as a lump sum measure of the antibody in residual plasma, interstitial fluid and cells. In terms of correlating antibody exposure to a therapeutic effect, however, interstitial pharmacokinetics might be more
Th M Behr et al.
Nuklearmedizin. Nuclear medicine, 41(2), 71-79 (2002-05-07)
Simple and reliable methodologies for radioiodination of proteins and peptides are described. The labeling systems are easy to assemble, capable of radioiodinating any protein or, with slight modifications, also peptide (molecular mass 1000-300,000) from kBq to GBq levels of activity
Iodination of chemokines for use in receptor binding analysis.
G L Bennett et al.
Methods in enzymology, 288, 134-148 (1997-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service