Skip to Content
Merck
All Photos(2)

Key Documents

SRP5096

Sigma-Aldrich

ULK1 (1-649), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ATG1, FLJ38455, UNC51, Unc51.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

46-62 nmol/min·mg

mol wt

~125 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ULK1(8408)

General description

Unc-51 like autophagy activating kinase 1 (ULK1) is a serine/threonine protein kinase which forms a component of the ULK1 protein kinase complex. This protein complex also contains the adaptor proteins ATG (autophagy-related) 13, RB1CC1/FIP200 (RB1-inducible coiled-coil 1), and ATG101. The gene encoding this protein is localized on human chromosome 12q24.33.

Application

GST-ULK1 has been used in in vitro kinase assay.

Biochem/physiol Actions

Unc-51 like autophagy activating kinase 1 (ULK1) protein kinase plays a crucial role in the control of autophagy initiation. This protein is responsible for activating a cascade of signaling pathways which results in the formation of phagosomes. ULK1 is also required for the control of the expression of key interferon-stimulated genes (ISGs).

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Computational analysis of an autophagy/translation switch based on mutual inhibition of MTORC1 and ULK1.
Szymanska Pp
PLoS ONE (2015)
Deubiquitinase inhibition by WP1130 leads to ULK1 aggregation and blockade of autophagy.
Driebren S
Autophagy (2015)
Deubiquitinase inhibition by WP1130 leads to ULK1 aggregation and blockade of autophagy
Stefan D
Autophagy (2015)
Missense Mutation in Pseudouridine Synthase 1 (PUS1) Causes Mitochondrial Myopathy and Sideroblastic Anemia (MLASA)
YelenaBykhovskaya
The American Journal on Addictions (2004)
Central role of ULK1 in type I interferon signaling.
Saleiro D
Cell Reports (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service