Skip to Content
Merck
All Photos(2)

Key Documents

SRP0415

Sigma-Aldrich

PKM1 active human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

OIP3, PK3, PKM2, TCB, pyruvate kinase, muscle

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

59 kDa

packaging

pkg of 20 μg

manufacturer/tradename

Sigma-Aldrich

storage condition

avoid repeated freeze/thaw cycles

concentration

1.8 mg/mL

technique(s)

cell based assay: suitable

solubility

water: soluble

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PKM(5315)

General description

Research area: Cell Signaling

Pyruvate kinase (PK) has four isozymes such as PKL, PKR, PKM1, and PKM2. PKM1/2 is located in the distal convoluted tubule of the kidney, embryo, lungs, proliferative tissues, retina, adipose tissues, and islets. The PKM1 gene is located on the human chromosome at 15q23.

Application

PKM1active human has been used in:
  • cell-based kinase assay to study the ability of a compound to inhibit the kinase activity of PKM1.
  • oxidative stress experiments on PKM2.
  • Co-immunoprecipitation (Co-IP) to determine pyruvate kinase M2 (PKM2) as a novel direct binding partner of phosphoserine aminotransferase 1 (PSAT1).

Biochem/physiol Actions

Pyruvate kinase (PK) catalyzes the dephosphorylation of phosphoenolpyruvate(PEP) into pyruvate. It is also involved in glycolysis by releasing ATP.Allosteric effects and intracellular signal transduction regulate the enzymaticactivity of PKM1. PKM1 is involved in several biological processes such asmetabolic reprogramming, and innate and acquired immune response. Itparticipates in protein interactions, protein synthesis, glycolysis, andintranuclear signal transduction.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear Pyruvate Kinase M2 (PKM2) Contributes to Phosphoserine Aminotransferase 1 (PSAT1)-Mediated Cell Migration in EGFR-Activated Lung Cancer Cells
Biyik-Sit R, et al.
Cancer, 13(16), 3938-3938 (2021)
C Parkison et al.
Biochemical and biophysical research communications, 179(1), 668-674 (1991-08-30)
Using a T7 expression system, the monomer of rat pituitary pyruvate kinase, subtype M1 (PKM1), was overexpressed in Escherichia coli and purified to homogeneity. The monomeric p58-M1 has intrinsic enzymatic activity with a Vmax of 79 +/- 20 units/mg and
Research progress on the role of PKM2 in the immune response
Chunyan L, et al.
Frontiers in Immunology (2022)
Development of Novel Irreversible Pyruvate Kinase M2 Inhibitors
I-Shan H, et al.
Journal of Medicinal Chemistry, 62(18), 8497-8510 (2019)
Evaluation of serum and pleural levels tumor M2-pyruvate kinase in lung cancer patients with pleural effusion
Tiantian Z, et al.
BMC Pulmonary Medicine, 22, 307-307 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service