Skip to Content
Merck
All Photos(2)

Key Documents

SPC25120

Sigma-Aldrich

SP Sephadex®

Synonym(s):

Sulfopropyl Sephadex®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

capacity

2.0-2.6 meq/g

Quality Level

General description

SPC25120-100G′s updated product number is GE17-0230-01

Application

SP Sephadex® is used in protein chromatography, ion exchange chromatography and cation exchange media. SP Sephadex® has been used to study human tumors as well as to discuss the biochemical basis for the use of mebendazole in the treatment of echinococcosis.

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and Characterization of Antioxidative Peptides from Protein Hydrolysate of Lecithin-Free Egg Yolk.
Park, P.-J., et al.
Journal of the American Oil Chemists' Society, 78, 651-656 (2001)
A W Mudge et al.
Proceedings of the National Academy of Sciences of the United States of America, 76(1), 526-530 (1979-01-01)
Sensory neurons grown in dispersed cell culture in the absence of non-neuronal cell types contain immunoreactive substance P that is chemically similar to synthetic substance P. When depolarized in high-K+ media (30-120 mM), the neurons release this peptide by a
J O Previato et al.
The Journal of biological chemistry, 273(24), 14982-14988 (1998-06-17)
In this study, we have characterized the activity of a uridine diphospho-N-acetylglucosamine:polypeptide-alpha-N-acetylglucosaminylt ransferase (O-alpha-GlcNAc-transferase) from Trypanosoma cruzi. The activity is present in microsomal membranes and is responsible for the addition of O-linked alpha-N-acetylglucosamine to cell surface proteins. This preparation adds
B Wurster et al.
Proceedings of the National Academy of Sciences of the United States of America, 73(3), 795-799 (1976-03-01)
Some species of cellular slime mold do not respond to cyclic AMP as an acrasin, or chemoattractant. In one such species, Polysphondylium violaceum, we have isolated and purified its acrasin and determined some of its chemical properties, which lead us
J F Kolhouse et al.
The Journal of biological chemistry, 266(34), 23010-23015 (1991-12-05)
Methionine synthase catalyzes the conversion of N5-methyltetrahydrofolate and homocysteine to tetrahydrofolate and methionine. Methylcobalamin (Me-Cbl) is tightly bound to methionine synthase and is required for enzymatic activity. When added to crude tissue homogenates, Me-Cbl stimulates methionine synthase but similar stimulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service