Skip to Content
Merck
All Photos(1)

Key Documents

SML3702

Sigma-Aldrich

Testosterone cypionate

≥98% (HPLC)

Synonym(s):

(17β)-17-(3-Cyclopentyl-1-oxopropoxy)androst-4-en-3-one, 17β-Hydroxyandrost-4-en-3-one cyclopentylpropionate, Cyclopentanepropionic acid ester with testosterone, NSC 9157

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40O3
CAS Number:
Molecular Weight:
412.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule III

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Orally available prodrug of testosterone that has a long elimination half-life; androgen that acts as agonist of the androgen receptor (AR)
Testosterone cypionate is a prodrug of testosterone that has a long elimination half-life. It is an androgen and anabolic–androgenic steroid that acts as agonist of the androgen receptor (AR). Testosterone cypionate is mainly used in the treatment of low testosterone levels in men. Also, it was approved for treatment of hypogonadism.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacology of testosterone replacement therapy preparations
Translational andrology and urology, 5(6), 834-843 (2016)
Durable Response of Enzalutamide-resistant Prostate Cancer to Supraphysiological Testosterone Is Associated with a Multifaceted Growth Suppression and Impaired DNA Damage Response Transcriptomic Program in Patient-derived Xenografts
European Urology, 77(2), 144-155 (2020)
Modulation of inflammatory and hormonal parameters in response to testosterone therapy: Effects on the ventral prostate of adult rats
Cell Biology International, 42(9), 1200-1211 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service