Skip to Content
Merck
All Photos(1)

Key Documents

SML3692

Sigma-Aldrich

SSR149415

≥98% (HPLC)

Synonym(s):

(-)-(2S,4R)-1-[5-Chloro-1-[(2,4-dimethoxyphenyl)sulphonyl]-3-(2-methoxyphenyl)-2-oxo-2,3-dihydro-1H-indol-3-yl]-4-hydroxy-N,N-dimethyl-2-pyrrolidine-carboxamide, (2S,4R)-1-[(3R)-5-Chloro-1-[(2,4-dimethoxyphenyl)sulfonyl]-2,3-dihydro-3-(2-methoxyphenyl)-2-oxo-1H-indol-3-yl]-4-hydroxy-N,N-dimethyl-2-pyrrolidinecarboxamide, Nelivaptan, SSR 149415, SSR-149415

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H32ClN3O8S
CAS Number:
Molecular Weight:
630.11
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

SSR149415 is an orally active, selective vasopressin V1b receptor antagonist (human/rat Ki in nM = 1.5/1.3/V1b, 91/1050/V1a, 1412/2897/V2, 174/270/OT) that inhibits 30 nM AVP-induced Ca2+ response in human and rat V1b CHO transfectants (Ki = 1.26/2.0 nM). SSR149415 suppresses AVP-mediated physiological responses in vivo, including corticotropin secretion (1-30 mg/kg p.o. or i.p. in rats) upon exogenous AVP administration (0.3 µg/kg alone or 0.03 µg/kg with 0.1 µg corticoliberin/kg via i.v.), restraint stress-Induced corticotropin secretion (EC50 = 10 mg/kg i.p. in rats), and in a murine model of anxiety (four-plate test; 1-10 mg/kg i.p. or 3-10 mg/kg p.o. acute or 10 mg/kg/day p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V1b Receptor Antagonist SSR149415 and Naltrexone Synergistically Decrease Excessive Alcohol Drinking in Male and Female Mice
Alcohol, clinical & experimental research, 42(1), 195-205 (2018)
Characterization of (2S,4R)-1-[5-chloro-1-[(2,4-dimethoxyphenyl)sulfonyl]-3-(2-methoxy-phenyl)-2-oxo-2,3-dihydro-1H-indol-3-yl]-4-hydroxy-N,N-dimethyl-2-pyrrolidine carboxamide (SSR149415), a selective and orally active vasopressin V1b receptor antagonist
Journal of Pharmacology and Experimental Therapeutics, 300(3), 1122-1130 (2002)
Differential activation of arginine-vasopressin receptor subtypes in the amygdaloid modulation of anxiety in the rat by arginine-vasopressin
Psychopharmacology, 235(4), 1015-1027 (2018)
Anxiolytic- and antidepressant-like effects of the non-peptide vasopressin V1b receptor antagonist, SSR149415, suggest an innovative approach for the treatment of stress-related disorders
Proceedings of the National Academy of Sciences of the USA, 99(9), 6370-6375 (2002)
Identification of avian vasotocin receptor subtype-specific antagonists involved in the stress response of the chicken, Gallus gallus
Journal of Biomolecular Structure & Dynamics, 37(7), 1685-1699 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service