Skip to Content
Merck
All Photos(2)

Key Documents

SML3656

Sigma-Aldrich

PAPTP trifluoroacetate

≥98% (HPLC)

Synonym(s):

(3-(4-(4-((7-Oxo-7H-furo[3,2-g]benzopyran-4-yl)oxy)butoxy)phenyl)propyl)triphenyl phosphonium trifluoroacetate, (3-(4-(4-(7-Oxo-7H-furo[3,2-g]chromen-4-yloxy)butoxy)phenyl)propyl)triphenylphosphonium trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H38O5P · xC2HF3O2
Molecular Weight:
653.72 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

storage temp.

-10 to -25°C

SMILES string

O=C1C=CC2=C(C3=C(C=C2O1)OC=C3)OCCCCOC4=CC=C(C=C4)CCC[P+](C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7

InChI

1S/C42H38O5P/c43-41-25-24-37-40(47-41)31-39-38(26-29-45-39)42(37)46-28-11-10-27-44-33-22-20-32(21-23-33)13-12-30-48(34-14-4-1-5-15-34,35-16-6-2-7-17-35)36-18-8-3-9-19-36/h1-9,14-26,29,31H,10-13,27-28,30H2/q+1

InChI key

MHIFCBNYOHOSHK-UHFFFAOYSA-N

Biochem/physiol Actions

PAPTP is a PAP-1-derivatized Kv1.3-selective potassium channel blocker with a positively charged lipophilic propyl-triphenylphosphonium (TP) moiety that allows mitochondria-targeted PAPTP delivery. Mitochondria Kv1.3 inhibition induces oxygen species (ROS)-mediated cancer-selective killing both in cultures (by 28%/69%/95% post 24-hr 0/1/10 µM PAPTP treatment of primary B-CLL; 20%/24% normal B-cell death with 0/20 µM PAPTP) and in murine orthotopic models of melanoma and pancreatic ductal adenocarcinoma in vivo (5 µmol/kg q.o.d. via i.p.). PAPTP exhibits higher anti-cancer efficacy than PAP-1 both in vitro and in vivo, and and is less affected by ultidrug resistance (MDR).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisa Venturini et al.
Neuro-Signals, 25(1), 26-38 (2017-09-05)
Glioblastoma (GBM) is one of the most aggressive cancers, counting for a high number of the newly diagnosed patients with central nervous system (CNS) cancers in the United States and Europe. Major features of GBM include aggressive and invasive growth
Sofia Parrasia et al.
Pharmaceuticals (Basel, Switzerland), 14(2) (2021-02-11)
A developing family of chemotherapeutics-derived from 5-(4-phenoxybutoxy)psoralen (PAP-1)-target mitochondrial potassium channel mtKv1.3 to selectively induce oxidative stress and death of diseased cells. The key to their effectiveness is the presence of a positively charged triphenylphosphonium group which drives their accumulation
Faye L Styles et al.
Cell death & disease, 12(4), 372-372 (2021-04-09)
Cellular energy metabolism is fundamental for all biological functions. Cellular proliferation requires extensive metabolic reprogramming and has a high energy demand. The Kv1.3 voltage-gated potassium channel drives cellular proliferation. Kv1.3 channels localise to mitochondria. Using high-resolution respirometry, we show Kv1.3
Luigi Leanza et al.
Cancer cell, 31(4), 516-531 (2017-04-12)
The potassium channel Kv1.3 is highly expressed in the mitochondria of various cancerous cells. Here we show that direct inhibition of Kv1.3 using two mitochondria-targeted inhibitors alters mitochondrial function and leads to reactive oxygen species (ROS)-mediated death of even chemoresistant
Roberta Peruzzo et al.
Frontiers in oncology, 7, 239-239 (2017-10-17)
Previous results link the mitochondrial potassium channel Kv1.3 (mitoKv1.3) to the regulation of apoptosis. By synthesizing new, mitochondria-targeted derivatives (PAPTP and PCARBTP) of PAP-1, a specific membrane-permeant Kv1.3 inhibitor, we have recently provided evidence that both drugs acting on mitoKv1.3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service