Skip to Content
Merck
All Photos(1)

Key Documents

SML3571

Sigma-Aldrich

GS-621763

≥98% (HPLC)

Synonym(s):

(2R,3R,4R,5R)-2-(4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-2-cyano-5-((isobutyryloxy)methyl)tetrahydrofuran-3,4-diyl bis(2-methylpropanoate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31N5O7
CAS Number:
Molecular Weight:
501.53
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#C[C@@]1(C2=CC=C3C(N)=NC=NN32)O[C@@H]([C@H]([C@H]1OC(C(C)C)=O)OC(C(C)C)=O)COC(C(C)C)=O

InChI

1S/C24H31N5O7/c1-12(2)21(30)33-9-16-18(34-22(31)13(3)4)19(35-23(32)14(5)6)24(10-25,36-16)17-8-7-15-20(26)27-11-28-29(15)17/h7-8,11-14,16,18-19H,9H2,1-6H3,(H2,26,27,28)/t16-,18-,19-,24+/m1/s1

InChI key

RVSSLHFYCSUAHY-JQGROFRJSA-N

Biochem/physiol Actions

GS-621763 is an orally bioavailable analog of remdesivir, which is efficiently converted in plasma to active metabolite GS-441524, or in lungs to the bioactive triphosphate GS-443902. GS-621763 effectively reduces SARS-CoV-2 burden to near-undetectable levels in ferrets.
Orally bioavailable analog of remdesivir, which is efficiently converted in plasma to active metabolite GS-441524

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therapeutic treatment with an oral prodrug of the remdesivir parental nucleoside is protective against SARS-CoV-2 pathogenesis in mice
Science Translational Medicine, 14 (2022)
Prodrugs of a 1'-CN-4-Aza-7,9-dideazaadenosine C-Nucleoside Leading to the Discovery of Remdesivir (GS-5734) as a Potent Inhibitor of Respiratory Syncytial Virus with Efficacy in the African Green Monkey Model of RSV
Journal of Medicinal Chemistry, 64, 5001-5017 (2021)
Oral prodrug of remdesivir parent GS-441524 is efficacious against SARS-CoV-2 in ferrets
Nature Communications, 12, 6415-6415 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service