Skip to Content
Merck
All Photos(1)

Key Documents

SML3230

Sigma-Aldrich

VU0360172 hydrochloride

≥98% (HPLC)

Synonym(s):

N-cyclobutyl-6-((3-fluorophenyl)ethynyl)nicotinamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15FN2O · HCl
CAS Number:
Molecular Weight:
330.78
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

FC1=CC=CC(C#CC2=NC=C(C=C2)C(NC3CCC3)=O)=C1.Cl

InChI

1S/C18H15FN2O.ClH/c19-15-4-1-3-13(11-15)7-9-16-10-8-14(12-20-16)18(22)21-17-5-2-6-17;/h1,3-4,8,10-12,17H,2,5-6H2,(H,21,22);1H

InChI key

NBGAPTWZQXSEAA-UHFFFAOYSA-N

General description

VU0360172 is a potent (EC50 = 16.6 nM), systemically active positive allosteric modulator (PAM) of the metabotropic glutamate receptor subtype 5 (mGlu5). It is the first orally active mGlu5 PAM to be efficacious in an in vivo preclinical antipsychotic model, producing a dose-dependent reversal of amphetamine-induced hyperlocomotion. It was also found to reduce spontaneous absence seizures in WAG/Rij rats and to inhibit pain. Reduced spontaneous spike and wave discharges without impacting motor behavior in a rat model of absence epilepsy have been observed.

Biochem/physiol Actions

VU0360172 is a potent (EC50 = 16.6 nM), systemically active positive allosteric modulator (PAM) of the metabotropic glutamate receptor subtype 5 (mGlu5). It is the first orally active mGlu5 PAM to be efficacious in an in vivo preclinical antipsychotic model, producing a dose-dependent reversal of amphetamine-induced hyperlocomotion. It was also found to reduce absence seizures in WAG/Rij rats and to inhibit pain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiation of mGlu5 receptors with the novel enhancer, VU0360172, reduces spontaneous absence seizures in WAG/Rij rats
V. D'Amore, et al.
Neuropharmacology, 330-338 (2013)
Alice L Rodriguez et al.
Molecular pharmacology, 78(6), 1105-1123 (2010-10-07)
Modulators of metabotropic glutamate receptor subtype 5 (mGluR5) may provide novel treatments for multiple central nervous system (CNS) disorders, including anxiety and schizophrenia. Although compounds have been developed to better understand the physiological roles of mGluR5 and potential usefulness for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service