Skip to Content
Merck
All Photos(1)

Key Documents

SML3206

Sigma-Aldrich

Ginsenoside Rg5

≥98% (HPLC)

Synonym(s):

(3β, 12β,20E)-12-Hydroxydammara-20(22),24-dien-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O12
CAS Number:
Molecular Weight:
767.00
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@H]([C@@H]2O)O)CO)O[C@@H]3C([C@H]4[C@@]([C@@H]5[C@]([C@]6([C@H]([C@@H](C5)O)[C@H](CC6)\C(=C\CC=C(C)C)\C)C)(CC4)C)(CC3)C)(C)C

InChI

1S/C42H70O12/c1-21(2)10-9-11-22(3)23-12-16-42(8)30(23)24(45)18-28-40(6)15-14-29(39(4,5)27(40)13-17-41(28,42)7)53-38-36(34(49)32(47)26(20-44)52-38)54-37-35(50)33(48)31(46)25(19-43)51-37/h10-11,23-38,43-50H,9,12-20H2,1-8H3/b22-11+/t23-,24-,25-,26-,27+,28-,2

InChI key

NJUXRKMKOFXMRX-RNCAKNGISA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yange Wei et al.
Journal of agricultural and food chemistry, 68(18), 5107-5117 (2020-04-21)
In this study, we aimed to investigate the influence of ginsenoside Rg5 (Rg5) on gut microbiota in diabetic db/db mice. Our data indicated that Rg5 not only improved the symptoms of hyperglycemia, repaired intestinal barrier function, and relieved metabolic endotoxemia-related
Yu Young Lee et al.
International journal of molecular sciences, 14(5), 9820-9833 (2013-05-24)
Microglia are resident immune cells in the central nervous system. They play a role in normal brain development and neuronal recovery. However, overactivation of microglia causes neuronal death, which is associated with neurodegenerative diseases, such as Parkinson's disease and Alzheimer's
Young-Lai Cho et al.
The Journal of biological chemistry, 290(1), 467-477 (2014-11-14)
Ginsenoside Rg5 is a compound newly synthesized during the steaming process of ginseng; however, its biological activity has not been elucidated with regard to endothelial function. We found that Rg5 stimulated in vitro angiogenesis of human endothelial cells, consistent with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service