Skip to Content
Merck
All Photos(1)

Key Documents

SML2980

Sigma-Aldrich

G6PDi-1

≥98% (HPLC)

Synonym(s):

4-([5-oxo-6H,7H,8H,9H-cyclohepta[d]pyrimidin-2-yl]amino)thiophene-2-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N4OS
CAS Number:
Molecular Weight:
284.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1=CC(NC2=NC=C3C(CCCCC3=N2)=O)=CS1

InChI

1S/C14H12N4OS/c15-6-10-5-9(8-20-10)17-14-16-7-11-12(18-14)3-1-2-4-13(11)19/h5,7-8H,1-4H2,(H,16,17,18)

InChI key

USWCHVCZOGGQQA-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

G6PDi-1 is a cell penetrant potent and selective nonsteroidal inhibitor of glucose-6-phosphate dehydrogenase (G6PD). G6PDi-1 strongly depletes NADPH in lymphocytes. It decreases inflammatory cytokine response in macrophages, and suppresses respiratory burst in neutrophils.
cell penetrant potent and selective nonsteroidal inhibitor of glucose-6-phosphate dehydrogenase (G6PD)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan M Ghergurovich et al.
Nature chemical biology, 16(7), 731-739 (2020-05-13)
Glucose is catabolized by two fundamental pathways, glycolysis to make ATP and the oxidative pentose phosphate pathway to make reduced nicotinamide adenine dinucleotide phosphate (NADPH). The first step of the oxidative pentose phosphate pathway is catalyzed by the enzyme glucose-6-phosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service