Skip to Content
Merck
All Photos(1)

Documents

SML2898

Sigma-Aldrich

Rupatadine fumarate

≥98% (HPLC)

Synonym(s):

8-Chloro-11-{1-[(5-methyl-3-pyridinyl)methyl]-4-piperidinylidene}-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine (E)-2-butenedioate (1:1), 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 8-chloro-6,11-dihydro-11-[1-[(5-methyl-3-pyridinyl)methyl]-4-piperidinylidene]-, (E)-2-butenedioate (1:1), UR 12592 fumarate, UR-12592 fumarate, UR12592 fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26ClN3 · C4H4O4
CAS Number:
Molecular Weight:
532.03
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C26H26ClN3.C4H4O4/c1-18-13-19(16-28-15-18)17-30-11-8-20(9-12-30)25-24-7-6-23(27)14-22(24)5-4-21-3-2-10-29-26(21)25;5-3(6)1-2-4(7)8/h2-3,6-7,10,13-16H,4-5,8-9,11-12,17H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+

InChI key

JYBLCDXVHQWMSU-WLHGVMLRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Rupatadine (UR-12592) is an orally active, dual histamine H1 and platelet-activating factor (PAF) receptor antagonist (Ki(app) = 100/550 nM against pyrilamine/WEB2086 binding to guinea pig cerebellum/rabbit platelet membranes) that inhibits histamine-induced guinea pig ileum contraction (pA2 = 9.29) and PAF-induced platelet aggregation (IC50 = 0.68 μM/human PRP). Rupatadine blocks histamine-/PAF-induced hypotension (ID50 = 1.4/0.44 mg/kg i.v. rats) and bronchoconstriction (ID50 = 113/9.6 μg/kg i.v. guinea pigs), as well as prevents mortality caused by PAF (ID50 = 0.31 mg/kg i.v. or 3.0 mg/kg p.o. mice) and endotoxin (mice/rats ID50 = 1.6/0.66 mg/kg i.v.) in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-xi Lv et al.
PloS one, 8(7), e68631-e68631 (2013-07-23)
A similar immune response is implicated in the pathogenesis of pulmonary fibrosis and allergic disorders. We investigated the potential therapeutic efficacy and mechanism of rupatadine, a dual antagonist of histamine and platelet-activation factor (PAF), in bleomycin- (BLM-) and silica-induced pulmonary
R Munoz-Cano et al.
Journal of investigational allergology & clinical immunology, 27(3), 161-168 (2016-10-21)
Platelet-activating factor (PAF) is a lipid mediator involved in the pathophysiology of several allergic diseases, for example, in the amplification of mast cell (MC) activation in anaphylaxis. Rupatadine is an antihistamine with a demonstrated anti-PAF effect, although its capacity to
Gathsaurie Neelika Malavige et al.
Scientific reports, 8(1), 3857-3857 (2018-03-03)
Currently there are no specific treatments available for acute dengue infection. We considered that rupatadine, a platelet-activating factor receptor inhibitor, might modulate dengue-associated vascular leak. The effects of rupatadine were assessed in vitro, and in a dengue model, which showed
R Caballero et al.
British journal of pharmacology, 128(5), 1071-1081 (1999-11-11)
1. The effects of rupatadine, a new dual antagonist of both histamine H1 and platelet-activating factor receptors, were studied on human cloned hKv1.5 channels expressed in Ltk- cells using the whole-cell patch-clamp technique. 2. Rupatadine produced a use- and concentration-dependent
Heba M Hafez et al.
Naunyn-Schmiedeberg's archives of pharmacology, 393(8), 1487-1500 (2020-03-23)
The role of histamine and platelet activating factor (PAF) as involved mediators in the pathophysiology of diabetic complications, in particular diabetic nephropathy (DN), has become a new focus of concern. Accordingly, the present study designed to explore the effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service