Skip to Content
Merck
All Photos(1)

Key Documents

SML2802

Sigma-Aldrich

ATR-101

≥98% (HPLC)

Synonym(s):

1[[1[4(Dimethylamino)phenyl]cyclopentyl]methyl]3[2,6di(propan2yl)phenyl]urea hydrochloride, ATR 101, ATR101, CI-984, N-[2,6-bis(1-Methylethyl)phenyl]-N-[[1-[4-(dimethylamino)phenyl]cyclopentyl]methyl]urea hydrochloride, Nevanimibe HCl, PD 132301 HCl, PD 132301-02, PD 132301-2, PD132301 HCl, PD132301-02, PD132301-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39N3O·HCl
CAS Number:
Molecular Weight:
458.08
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Cl-].N(C)(C)c1ccc(cc1)C3(CCCC3)CNC(=O)Nc2c(cccc2C(C)C)C(C)C.[H+]

InChI

1S/C27H39N3O.ClH/c1-19(2)23-10-9-11-24(20(3)4)25(23)29-26(31)28-18-27(16-7-8-17-27)21-12-14-22(15-13-21)30(5)6;/h9-15,19-20H,7-8,16-18H2,1-6H3,(H2,28,29,31);1H

InChI key

SDOOGTHIDFZUNM-UHFFFAOYSA-N

Biochem/physiol Actions

ATR-101 (PD 132301-02; Nevanimibe HCl) is an orally active, potent and selective acyl-CoA:cholesterol acyltransferase 1 (ACAT1, SOAT1) inhibtior (IC50 = 52 nM; intestinal microsome from cholesterol-fed rabbits) that reduces plasma cholesterol in both acute (by 77%; 50 mg/kg po.) and chronic (by 80%; 10 mg/kg po.) cholesterol-fed rat models. In addition, ATR-101 is reported to exhibit therapeutic efficacy against adrenocortical carcinoma (ACC), congenital adrenal hyperplasia (CAH), and Cushing′s syndrome (CS).
Orally active, potent and selective acyl-CoA:cholesterol acyltransferase 1 (ACAT1, SOAT1) inhibtior.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunhui Cheng et al.
Endocrine-related cancer, 23(4), 1-19 (2016-02-05)
Adrenocortical carcinoma (ACC) generally has poor prognosis. Existing treatments provide limited benefit for most patients with locally advanced or metastatic tumors. We investigated the mechanisms for the cytotoxicity, xenograft suppression, and adrenalytic activity of ATR-101 (PD132301-02), a prospective agent for
J F Reindel et al.
Toxicologic pathology, 22(5), 510-518 (1994-09-01)
PD 132301-2, an acyl-CoA: cholesterol acyltransferase (ACAT) inhibitor, was administered orally to cynomolgus monkeys for 2 wk at doses of 25, 50, 100, and 200 mg/kg to assess potential subacute toxicity. Sporadic episodes of soft feces and diarrhea increased in
M A Dominick et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 20(2), 217-224 (1993-02-01)
PD 132301-2 is a substituted urea hypolipidemic and antiatherosclerotic agent that is a potent inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT). To determine its subacute toxicity, PD 132301-2 was administered orally to beagle dogs at 0, 6, 12, 25, 50, 200, 400
B K Trivedi et al.
Journal of medicinal chemistry, 37(11), 1652-1659 (1994-05-27)
We recently described our initial structure-activity relationship (SAR) studies on a series of N-phenyl-N'-aralkyl- and N-phenyl-N'-(1-phenylcycloalkyl)ureas as inhibitors of acyl-CoA: cholesterol acyltransferase (ACAT). From this series of analogs, compound 1 (PD 129337) was identified as a potent inhibitor of ACAT
R G Elkin et al.
Poultry science, 72(3), 513-520 (1993-03-01)
PD132301-2, an inhibitor of acyl-coenzyme A:cholesterol O-acyltransferase (ACAT; EC 2.3.1.26), and 1-stearylboronic acid (SBA), a fatty acid analogue, were orally administered to White Leghorn hens in separate experiments to evaluate their effects on layer performance and plasma and egg yolk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service