Skip to Content
Merck
All Photos(1)

Key Documents

SML2652

Sigma-Aldrich

Peretinoin

≥97% (HPLC)

Synonym(s):

3,7,11,15-Tetramethyl-2,4,6,10,14-hexadecapentaenoic acid, (2E,4E,6E,10E)-3,7,11,15-Tetramethylhexadeca-2,4,6,10,14-pentaenoic acid, 4,5-Didehydrogeranyl geranoic acid, ACR, Acyclic retinoid, E 5166, E-5166, E5166, K 333, K-333, K333, NIK 333, NIK-333, NIK333, Polyprenoic acid, all-trans-3,7,11,15-Tetramethyl-2,4,6,10,14-hexadecapentaenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C20H30O2/c1-16(2)9-6-10-17(3)11-7-12-18(4)13-8-14-19(5)15-20(21)22/h8-9,11,13-15H,6-7,10,12H2,1-5H3,(H,21,22)/b14-8+,17-11+,18-13+,19-15+

InChI key

UUBHZHZSIKRVIV-KCXSXWJSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Peretinoin is an orally active synthetic acyclic retinoid (ACR) whose in vivo anti-hepatocellular carcinoma (HCC) efficacy is attributed to its upregulatory activity toward retinoid nuclear receptors (RARbeta & RXRalpha), as well as negative regulation against sphingosine kinase 1 (SPHK1) expression and, thereby, sphingosine metabolic pathway. Peretinoin inhibits the replication of hepatitis B & C viruses in cultures (HBV & HCV EC50 = 9-25 μM) and shows in vivo efficacy in rodent models of hepatocarcinogenesis among rats (10, 40, or 80 mg/kg/day p.o.) and mice (0.03% or 0.06% in diet) subjected to chronic inflammation induction by diethylnitrosamine (DEN).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xian-Yang Qin et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(19), 4969-4974 (2018-04-25)
Hepatocellular carcinoma (HCC) is a highly lethal cancer that has a high rate of recurrence, in part because of cancer stem cell (CSC)-dependent field cancerization. Acyclic retinoid (ACR) is a synthetic vitamin A-like compound capable of preventing the recurrence of
Masaya Funaki et al.
Scientific reports, 7(1), 16978-16978 (2017-12-07)
Sphingosine-1-phospate is a potent bioactive lipid metabolite that regulates cancer progression. Because sphingosine kinase 1 and sphingosine kinase 2 (SPHK 1/2) are both essential for sphingosine-1-phospate production, they could be a therapeutic target in various cancers. Peretinoin, an acyclic retinoid
Tetsuro Sano et al.
Nutrition and cancer, 51(2), 197-206 (2005-04-30)
We investigated the preventive effects of a synthetic acyclic retinoid, NIK-333, on the early and late events of hepatocarcinogenesis in male F344 rats treated with 3'-methyl-4-dimethylaminoazobenzene (3'-MeDAB). NIK-333 was administered once a day on consecutive days at a dose of
Masataka Kagawa et al.
Carcinogenesis, 25(6), 979-985 (2004-01-27)
The present study was designed to determine the effects of NIK-333, a synthetic acyclic retinoid, on N-diethylnitrosamine (DEN)-induced hepatocarcinogenesis in male F344 rats. Animals were given DEN dissolved in drinking water at a concentration of 40 p.p.m. for 5 weeks
Y Muto et al.
The New England journal of medicine, 334(24), 1561-1567 (1996-06-13)
In patients with hepatocellular carcinoma (hepatoma), the rate of recurrent and second primary hepatomas is high despite surgical resection and percutaneous ethanol-injection therapy. We developed an acyclic retinoid, polyprenoic acid, that inhibits hepatocarcinogenesis in the laboratory and induces differentiation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service