Skip to Content
Merck
All Photos(1)

Key Documents

SML2646

Sigma-Aldrich

CX614

≥98% (HPLC)

Synonym(s):

2,3,6a,7,8,9-Hexahydro-11H-1,4-dioxino[2,3-g]pyrrolo[2,1-b][1,3]benzoxazin-11-one, 2H,3H,6aH-Pyrrolidino[2",1"-3′,2′]1,3-oxazino[6′,5′-5,4]benzo[e]1,4-dioxan-10-one, BDP 37, BDP-37, CX 614, CX-614, LiD 37

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO4
CAS Number:
Molecular Weight:
247.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N21C(Oc3c(cc4c(c3)OCCO4)C2=O)CCC1

InChI

1S/C13H13NO4/c15-13-8-6-10-11(17-5-4-16-10)7-9(8)18-12-2-1-3-14(12)13/h6-7,12H,1-5H2

InChI key

RQEPVMAYUINZRE-UHFFFAOYSA-N

Biochem/physiol Actions

CX614 (BDP-37) is a brain-penetrant class I ampakine (benzoxazine subgroup of benzamide-type) with AMPA receptor (AMPAR; iGluR) positive allosteric modulator (PAM) activity via targeting a binding site shared by cyclothiazide (CTZ), but distinct from that of GYKI 52466. CX614 enhances field excitatory postsynaptic potentials (amplitude & duration) in rat hippocampal slices and autaptically evoked excitatory postsynaptic currents in neuronal cultures (EC50 of 20-40 μM) by blocking desensitization and slowing deactivation of responses to glutamate. CX614 is also widely employed for studying AMPAR-mediated physiological responses in vivo (1-10 mg/kg n rats and mice via i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hennegriff et al.
Journal of neurochemistry, 68(6), 2424-2434 (1997-06-01)
Homomeric AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid)-type glutamate receptors (GluRs) were stably expressed in kidney cells from cDNAs encoding GluR1 flop, GluR2 flip, GluR2 flop, and GluR3 flop subunits. The recombinant receptors were of the expected size and showed functional properties in whole-cell
J C Lauterborn et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 20(1), 8-21 (2000-01-11)
This study investigated whether positive modulators of AMPA-type glutamate receptors influence neurotrophin expression by forebrain neurons. Treatments with the ampakine CX614 markedly and reversibly increased brain-derived neurotrophic factor (BDNF) mRNA and protein levels in cultured rat entorhinal/hippocampal slices. Acute effects
U S Hess et al.
Neuroscience, 121(2), 509-521 (2003-10-03)
It has been proposed that glutamatergic and dopaminergic systems are functionally opposed in their regulation of striatal output. The present study tested the effects of drugs that enhance AMPA-receptor-mediated glutamatergic transmission (ampakines) for their effects on dopamine-related alterations in cortical
Daniel P Radin et al.
Anticancer research, 38(6), 3461-3465 (2018-06-01)
Mounting evidence suggests that trophic cell signaling can be mediated by alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) activation. It has been demonstrated that exogenous application of brain-derived neurotrophic factor (BDNF) is highly neuroprotective in vitro against neurotoxic insults such as standard chemotherapies.
Daniel P Radin et al.
Biochemical pharmacology, 154, 446-451 (2018-06-16)
Transmembrane AMPA receptor regulatory proteins (TARPs) govern AMPA receptor cell surface expression and distinct physiological properties including agonist affinity, desensitization and deactivation kinetics. The prototypical TARP, STG or γ2 and TARPs γ3, γ4, γ7 and γ8 are all expressed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service