Skip to Content
Merck
All Photos(1)

Key Documents

SML2643

Sigma-Aldrich

ML365

≥98% (HPLC)

Synonym(s):

2-Methoxy-N-[3-[(3-methylbenzoyl)amino]phenyl]benzamide, ML 365, ML365

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2O3
CAS Number:
Molecular Weight:
360.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c2cc(ccc2)NC(=O)c3cc(ccc3)C)C(=O)c1c(cccc1)OC

InChI

1S/C22H20N2O3/c1-15-7-5-8-16(13-15)21(25)23-17-9-6-10-18(14-17)24-22(26)19-11-3-4-12-20(19)27-2/h3-14H,1-2H3,(H,23,25)(H,24,26)

InChI key

UTAJHKSGYJSZBR-UHFFFAOYSA-N

Biochem/physiol Actions

Orignially characterized as an mGluR5 negative allosteric modulator (IC50 = 1.35 μM, Emax = 3.82% of Glu Emax; rat mGluR5 HEK293A cells), ML365 is now better known as selective two-pore potassium (K2P) channel TASK1 (K2P3.1; KCNK3) inhibitor with 100- and 62-fold selectivity over TASK3 (K2p9.1; KCNK9), respectively, by thellium flux (rat TASK1/TASK3 CHO cell IC50 = 4/390 nM) and QPatch assay (rat TASK1/TASK3 CHO cell IC50 = 16/990 nM). ML365 is employed in the range from 40 nM to 20 μM. Selective TASK1 blockage is achieved at the lower end of the concentration range, while TASK3 activity can also be inhibited at the high end of the range.
Selective two-pore potassium (K2P) channel TASK1 (K2P3.1; KCNK3) inhibitor with 62- to 100-fold selectivity over TASK3 (K2p9.1; KCNK9).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas C Vierra et al.
Science signaling, 10(497) (2017-09-21)
Ca2+ handling by the endoplasmic reticulum (ER) serves critical roles in controlling pancreatic β cell function and becomes perturbed during the pathogenesis of diabetes. ER Ca2+ homeostasis is determined by ion movements across the ER membrane, including K+ flux through
Ya Zhou et al.
Bioorganic & medicinal chemistry letters, 19(23), 6502-6506 (2009-10-31)
This Letter describes the discovery and SAR of three novel series of mGluR5 non-competitive antagonists/negative allosteric modulators (NAMs) not based on manipulation of an MPEP/MTEP chemotype identified by a functional HTS approach. This work demonstrates fundamentally new mGluR5 NAM chemotypes
M A Skarsfeldt et al.
Acta physiologica (Oxford, England), 223(3), e13049-e13049 (2018-02-08)
The zebrafish has emerged as a novel model for investigating cardiac physiology and pathology. The aim of this study was to investigate the atrium-specific ion channels responsible for shaping the atrial cardiac action potential in zebrafish. Using quantitative polymerase chain
Daniel P Flaherty et al.
Bioorganic & medicinal chemistry letters, 24(16), 3968-3973 (2014-07-16)
TASK-1 is a two-pore domain potassium channel that is important to modulating cell excitability, most notably in the context of neuronal pathways. In order to leverage TASK-1 for therapeutic benefit, its physiological role needs better characterization; however, designing selective inhibitors
Yun-Fei Bai et al.
Biochemical and biophysical research communications, 503(1), 79-85 (2018-06-01)
The noradrenergic neurons of the locus coeruleus (LC) are associated with various brain functions and psychiatric disorders, such as addiction and depression. It has been shown that neuropeptide galanin (GAL) inhibits neuronal excitability in LC, but the mechanisms remain unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service