Skip to Content
Merck
All Photos(1)

Key Documents

SML2627

Sigma-Aldrich

Connexin43 mimetic peptide 5 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Peptide 5 trifluoroacetate salt, VDCFLSRPTEKT trifluoroacetate salt, Val-Asp-Cys-Phe-Leu-Ser-Arg-Pro-Thr-Glu-Lys-Thr trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H98N16O20S · xC2HF3O2
CAS Number:
Molecular Weight:
1395.58 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

lyophilized powder

color

white to beige

storage temp.

−20°C

Biochem/physiol Actions

Connexin43 mimetic peptide 5 is a blocker of connexin43 hemichannels that inhibits gap junction channels following CNS injury. Peptide 5 was designed to bind to extracellular regions of the connexin43 protein.
blocker of connexin43 hemichannels that inhibits gap junction channels following CNS injury

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yilin Mao et al.
Experimental brain research, 235(10), 3033-3048 (2017-07-21)
Systemic administration of a Connexin43 mimetic peptide, Peptide5, has been shown to reduce secondary tissue damage and improve functional recovery after spinal cord injury (SCI). This study investigated safety measures and potential off-target effects of Peptide5 systemic administration. Rats were
Helen V Danesh-Meyer et al.
Brain : a journal of neurology, 135(Pt 2), 506-520 (2012-02-22)
Connexin43 gap junction protein is expressed in astrocytes and the vascular endothelium in the central nervous system. It is upregulated following central nervous system injury and is recognized as playing an important role in modulating the extent of damage. Studies
Yeri Kim et al.
Biochimica et biophysica acta. General subjects, 1861(2), 68-78 (2016-11-07)
Non-selective Connexin43 hemichannels contribute to secondary lesion spread. The hemichannel blocking peptidomimetic Peptide5, derived from the second extracellular loop of the human Connexin43 protein, prevents lesion spread and reduces vascular permeability in preclinical models of central nervous system injury. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service